Maxima H Minus Reverse Transcriptase (200 U/μL), 2,000 Units - FAQs
Maxima H Minus Reverse Transcriptase (200 U/μL), 2,000 Units - FAQs
View additional product information for Maxima H Minus Reverse Transcriptase (200 U/μL) - FAQs (EP0751, EP0752, EP0753)
7 product FAQs found
When should I choose regular RevertAid RT or Maxima RT vs. RevertAid H-minus RT or Maxima H-minus RT?
It is generally beneficial to minimize RNase H activity when aiming to produce long transcripts for cDNA cloning. RNase H degrades RNA from RNA-DNA duplexes, which can result in truncated cDNA during reverse transcription of long mRNA. We also recommended using RNase H-minus RTs for template-independent addition of C nucleotides.
Is it preferable to use elevated temperature in the RT reaction when using Maxima reverse transcriptases?
cDNA synthesis at higher temperatures ensures successful transcription of RNA with high levels of secondary structure, reducing issues of primer access to template. Therefore, we do recommend to use RT enzymes with high thermostability, e.g. Maxima and Maxima H Minus Reverse Transcriptases, which provide higher yields of full-length cDNA, better sensitivity, and successful transcription of GC-rich templates.
What steps should I take while performing first strand cDNA synthesis using low purity template (e.g., inhibitors in RNA sample)?
Trace amounts of reagents used in RNA purification protocols may remain in solution and inhibit first-strand synthesis, e.g., SDS, EDTA, guanidine salts, phosphate, pyrophosphate, polyamines, spermidine. To remove trace contaminants, we recommend re-precipitating the RNA with ethanol and washing the pellet with 75% ethanol, or re-purifying the RNA.
For reverse transcription, how important is the quality of RNA template?
RNA purity and integrity are essential for synthesis and quantification of cDNA. Always assess the integrity of RNA prior to cDNA synthesis. Use freshly prepared RNA. Multiple freeze/thaw cycles of the RNA sample and synthesized cDNA is not recommended. Avoid RNase contamination and discard low quality RNA.
When should I choose regular RevertAid RT or Maxima RT vs. RevertAid H Minus RT or Maxima H Minus RT?
It is generally beneficial to minimize RNase H activity when aiming to produce long transcripts for cDNA cloning. RNase H degrades RNA from RNA-DNA duplexes, which can result in truncated cDNA during reverse transcription of long mRNA. We also recommended using RNase H Minus RTs for template-independent addition of C nucleotides. In contrast, reverse transcriptases with intrinsic RNase H activity are often favored in 1-step RT-qPCR applications.
Do Thermo Scientific reverse transcriptases (RevertAid RT, RevertAid H Minus RT, Maxima RT, and Maxima H Minus RT) possess terminal deoxynucleotidyl (TdT) activity?
All Thermo Scientific reverse transcriptases (RevertAid RT, RevertAid H Minus RT, Maxima RT, and Maxima H Minus RT) possess intrinsic TdT activity, although at varying degrees depending upon the reaction conditions. We recommend specialized SuperScript IV Template Switching RT Master Mix for high efficiency in applications requiring template switching RT.
Do your 'engineered' MMLV-derived reverse transcriptase enzymes (e.g., SuperScript IV RT and Maxima RT) retain the ability to add a few additional nucleotides to the 3' end of the newly synthesized cDNA strand?
Regarding the tailing/TdT activity, a recent comparative analysis by R&D has shown that SuperScript IV RT has a much better TdT activity than SuperScript III RT, and that the latter has lost almost all activity.
Regarding the length of fragments that can be synthesized, internal tests have shown that fragments of up to 13 kb can be synthesized using SuperScript IV RT. Larger fragments have not been tested yet. Maxima H- RT was successfully tested with fragments up to 20 kb and since it has a certain amount of TdT activity, this enzyme could also be an alternative.
For template switching, we would recommend using SuperScript IV RT in the first instance, but Maxima H- RT would also work.