RNA Polymerases
Thermo Scientific™

RNA Polymerases

Thermo Scientific enzymes for in vitro transcription are designed for efficient and reliable RNA synthesis. These enzymes cover the entire synthesis workflow from template preparation to post-transcriptional modifications, ensuring optimal performance and high yields of synthesized RNA.
製品番号(カタログ番号)ポリメラーゼ濃度数量
EP168AFポリ(A)RNAポリメラーゼ5 U/μL800 U
EP0111AFT7 RNAポリメラーゼ200 U/μL20,000 U
製品番号(カタログ番号) EP168AF
価格(JPY)
48,300
キャンペーン価格
Ends: 27-Mar-2026
80,500
割引額 32,200 (40%)
Each
ポリメラーゼ:
ポリ(A)RNAポリメラーゼ
濃度:
5 U/μL
数量:
800 U

Thermo Scientific enzymes for in vitro transcription are designed for efficient and reliable RNA synthesis. These enzymes cover the entire synthesis workflow from template preparation to post-transcriptional modifications, ensuring optimal performance and high yields of synthesized RNA.

High quality RNA synthesized by in vitro transcription is essential for RNA research and this versatile technique can be used in a variety of applications including the introduction of modified nucleotides into RNA transcripts for enhanced translational capacity and reduced immune activity of the cells.

Thermo Scientific enzymes for in vitro transcription are designed for efficient and reliable RNA synthesis. These enzymes cover the entire synthesis workflow from template preparation to post-transcriptional modifications, ensuring optimal performance and high yields of synthesized RNA.

High quality RNA synthesized by in vitro transcription is essential for RNA research and this versatile technique can be used in a variety of applications including the introduction of modified nucleotides into RNA transcripts for enhanced translational capacity and reduced immune activity of the cells.

Features

  • Convenient stand-alone enzymes and buffers for workflow optimization
  • No animal-origin components in final product composition
  • Reliable and reproducible results with rigorous quality testing
  • Scalable and available as OEM products

Applications

  • CRISPR/Cas-based genome editing
  • Gene expression
  • Generation of pluripotent cells
研究用途にのみご使用ください。診断目的には使用できません。
仕様
濃度5 U/μL
使用対象(アプリケーション)RNA Polyadenylation
数量800 U
ポリメラーゼポリ(A)RNAポリメラーゼ
製品ラインTheraPure
Unit SizeEach
組成および保存条件
5 U/μL Poly(A) Polymerase, 160 μL

Store at -20°C.