Mouse Detection Antibody Diluent
Invitrogen™

Mouse Detection Antibody Diluent

Antibody Dilution Buffer
Have Questions?
Catalog NumberQuantity
EPX-22222-0001 each
Catalog number EPX-22222-000
Price (EUR)
71,23
Online exclusive
92,50
Save 21,27
Each
Quantity:
1 each
Price (EUR)
71,23
Online exclusive
92,50
Save 21,27
Each
The ProcartaPlex Detection Antibody Diluent is used for dilution of detection antibodies.
The ProcartaPlex Detection Antibody Diluent is included in our basic kits and off the shelf panels. Additionally, we offer the Detection Antibody Diluent as a stand-alone item.

Reported Application
Multiplex Immunoassay

For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)Luminex™ Instruments
Product TypeDiluent
Quantity1 each
SpeciesMouse
Unit SizeEach

Frequently asked questions (FAQs)

Are the standalone ProcartaPlex assay buffers lot-specific such that they will only work with specific lots of other components?

No, the standalone ProcartaPlex assay buffers are not lot-specific.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

Can I get the catalog numbers for ProcartaPlex assay buffers and plates?

Here are the catalog numbers:

- EPX-11111-000: ProcartaPlex Universal Assay Buffer (1X) (10 mL) - It replaces all discontinued Procarta buffers (standard, assay, sample buffer); it is the same as the discontinued Procarta Cell Lysate Assay Buffer, 10 plate (Cat. No. PC0165), but only for one plate
- EPX-11110-000: ProcartaPlex Universal Assay Buffer (10X) (10 mL)
- EPXP-11112-000: ProcartaPlex Platinum Assay Buffer for Plasma Samples (1X, 10 mL)
- EPXP-11113-000: ProcartaPlex Platinum Assay Buffer for Serum Samples (1X, 10 mL)
- EPX-99999-000: ProcartaPlex Cell Lysis Buffer; It is the same as the discontinued Procarta Cell lysis Buffer (for cell lysate preparation) (Cat. No. PC6002)
- EPX-22222-000: ProcartaPlex Detection Antibody Diluent, Mouse (1X, 3 mL)
- EPX-33333-000: ProcartaPlex Detection Antibody Diluent, Human (1X, 3 mL)
- EPX-66666-000: ProcartaPlex Wash Buffer 10X (20 mL)
- EPX-66666-001: ProcartaPlex Wash Buffer 10X (200 mL)
- EPX-77777-000: ProcartaPlex Reading Buffer (1X, 40 mL)
- EPX-SAPE-000: ProcartaPlex Streptavidin-PE (1X, 5 mL) - 4.3 µg/mL (can range from 4.25-4.35 µg/mL)
- EPX-44444-000: ProcartaPlex 96 well Flat Bottom Plate, black (1 plate)
- EPX-88888-000: ProcartaPlex 96 well Filter Plate (1 plate)
- EPX-38488-000: ProcartaPlex 384 well Filter Plate (1 plate)

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.