SARS-CoV-2 Spike Omicron (B.1.1.529) Neutralizing Antibody Human ProcartaPlex™ Simplex Kit
Invitrogen™

SARS-CoV-2 Spike Omicron (B.1.1.529) Neutralizing Antibody Human ProcartaPlex™ Simplex Kit

The Human SARS-CoV-2 Spike Omicron (B.1.1.529) Neutralizing Antibody ProcartaPlex Simplex Kit enables detection of neutralizing anti-SARS-CoV-2 antibodies in serum and plasma.
Have Questions?
Número do catálogoQuantity
EPX010-16019N-90196 Tests
Número do catálogo EPX010-16019N-901
Preço (BRL)
-
Quantity:
96 Tests

The Human SARS-CoV-2 Spike Omicron (B.1.1.529) Neutralizing Antibody ProcartaPlex Simplex Kit enables detection of neutralizing anti-SARS-CoV-2 antibodies in serum and plasma. The assay enables measurement of the neutralizing potential of antibodies towards the variant B.1.1.529 (o) using Luminex xMAP technology.

Unlike most ProcartaPlex kits that utilize matched antibody pairs to detect proteins of interest, this serological assay is designed with SARS-CoV-2 S1 variant protein B.1.1.529 (o) conjugated to the bead. Samples or controls are added and antibodies bind to the conjugated proteins. This is a competitive assay where biotinylated ACE2 is used as detection reagent. Biotinylated ACE2 will bind to the spike protein on the beads when not blocked by bound neutralizing antibodies. Streptavidin-phycoerythrin (PE) is added and the signal is indirectly proportional to the amount of specific neutralizing antibodies present. Negative controls will give the highest signal, while neutralizing antibodies in the sample or the positive control will reduce the signal.

The total incubation time of the Human SARS-CoV-2 Spike Omicron (B.1.1.529) Neutralizing Antibody ProcartaPlex Simplex Kit is three hours. The kit comes with all reagents needed to run the assay (no need to purchase a basic kit) and includes two vials of positive controls. Serum and plasma samples are recommended to be diluted 1:100. This kit is not combinable with other simplex kits or panels.

Especificações
Assay RangeSee product documentation
Assay SensitivitySee Certificate of Analysis
Bead TypeMagnetic
For Use With (Equipment)Luminex™ Instruments
CombinabilityCombinable
Product TypeSimplex Kit
Quantity96 Tests
Research AreaSARS-CoV-2 Ig Total
SpeciesHuman
Unit SizeEach
Conteúdo e armazenamento

• 1 vial Standard Mix
• 1 vial Bead Mix (1X)
• 2 vials Control
• 1 vial Biotinylated Detection Antibody Mix (1X)
• 1 bottle Reading Buffer (1X)
• 1 bottle Wash Buffer (10X)
• 1 bottle Streptavidin-PE (1X)
• 1 bottle Universal Assay Buffer (1X)
• 1 bottle Detection Antibody Diluent (1X)
• 8-tube strip
• Adhesive film
• Flat bottom 96-well plate, black

Store at 2°C to 8°C.