p38 Human ProcartaPlex™ Simplex Kit
Invitrogen™
p38 Human ProcartaPlex™ Simplex Kit
Invitrogen ProcartaPlex Human p38 Simplex Kit enables precise analysis of the Protein of Interest.
Have Questions?
카탈로그 번호수량
EPX010-97010-90196 Tests
카탈로그 번호 EPX010-97010-901
제품 가격(KRW)
835,000
Each
수량:
96 Tests
제품 가격(KRW)
835,000
Each
The ProcartaPlex Human p38 Simplex Kit measures the level of total p38 in cell lysates. It is designed to be combinable with other Simplex kits so you can create your own multiplex panel that utilizes Luminex xMAP technology for protein quantitation. When combining multiple Simplex kits (i.e., when you are not using a pre-configured Multiplex Panel), only one buffer kit (sold separately) is needed for each assay plate regardless of plex size.
For preparation of cell lysates, we recommend the use of either ProcartaPlex Cell Lysis Buffer (Cat. No. EPX-99999-000) or Cell Extraction Buffer (Cat. No. FNN0011) supplemented with 0.5 M EDTA and Halt Protease and Phosphatase Inhibitor Single to Use Cocktail 100X (Cat. No. 78440 or 78442). These buffers are not included in the kit. Depending on your cell system, other buffers can be suitable as well.
The ProcartaPlex Human p38 Simplex Kit is part of the ProcartaPlex Human MAPK Pathway Panel (total) 6plex (Cat. No. EPX060-97017-901), which complements the ProcartaPlex Human MAPK Pathway Panel (phospho) 6plex (Cat. No. EPX060-97117-PHO).
About ProcartaPlex Assays for the Luminex Platform
ProcartaPlex immunoassays are based on the principles of a sandwich ELISA, using two highly specific antibodies binding to different epitopes of one protein to quantitate all protein targets simultaneously using a Luminex instrument. ProcartaPlex multiplex assays require as little as 25 μL of plasma or serum, or 50 μL of cell culture supernatant, and just four hours to obtain analyzed results.
  • More results per sample - measure multiple protein targets simultaneously in a single 25 to 50 μL sample
  • Well-established Luminex technology - highly referenced multiplexing platform for protein detection and quantitation
ProcartaPlex assays utilize Luminex xMAP (multianalyte profiling) technology for the simultaneous detection and quantitation of up to 80 protein targets in a single 25 to 50 μL sample from plasma, serum, cell culture supernatants, and other bodily fluids.
The Luminex beads in the ProcartaPlex assay are internally dyed with precise proportions of red and infrared fluorophores to create spectrally unique signatures that can be identified by the Luminex xMAP detection systems (e.g., Luminex 200, FLEXMAP 3D, and MAGPIX). Similar to a sandwich ELISA, the ProcartaPlex assay uses matched antibody pairs to identify the protein of interest. In a multiplexed assay, each spectrally unique bead is labeled with antibodies specific for a single target protein, and bound proteins are identified with biotinylated antibodies and streptavidin to R-phycoerythrin (RPE). The conjugation of protein-specific antibodies to a distinct bead allows for analysis of multiple targets in a single well.
The most significant difference between a ProcartaPlex assay and ELISA is that the capture antibody in the ProcartaPlex assay is conjugated to a bead and not adsorbed to the microplate well, so the ProcartaPlex assay reagents are free-floating in the solution. For detection, the Luminex 200 instrument, for example, contains two lasers, one to distinguish the spectral signature of each bead and the second to quantify the amount of RPE fluorescence, which is proportional to the amount of protein present in the sample. ProcartaPlex multiplex assays can profile more target proteins using significantly less sample in the same time that it takes to perform a traditional sandwich ELISA.
For Research Use Only.
사양
어세이 범위See Certificate of Analysis
어세이 민감성See Certificate of Analysis
비드 영역p38 [14]
비드 유형Magnetic
용도(장비)Luminex™ Instruments
유전자MAPK11
유전자 별칭PRKM11
유전자 ID(Entrez)5600
유전자 기호MAPK11
고처리량 호환성Yes
분석 간 CVSee Certificate of Analysis
분석 내 CVSee Certificate of Analysis
제품라인ProcartaPlex
샘플 종류Cell lysates
샘플 부피Cell lysates (prediluted): 25 μL
배송 조건Wet Ice
동의어p38 mitogen-activated protein kinase
UniProt IDQ15759
CombinabilityCombinable
제품 유형Simplex Kit
수량96 Tests
Research AreaSignaling
Human
Unit SizeEach
구성 및 보관
1 vial Capture Beads (50x)
1 vial biotinylated Detection Antibody (50x)
2 vials Standard Mix (lyophilized)
Store as indicated on the Certificate of Analysis.