Invitrogen™

IL-20 Human ProcartaPlex™ Simplex Kit

Catalog number: EPX01A-12168-901
Invitrogen™

IL-20 Human ProcartaPlex™ Simplex Kit

Catalog number: EPX01A-12168-901
Catalog Number
EPX01A-12168-901
Unit Size
96 tests
Price (USD)
Price: 302.00
Your price:
Quantity
Catalog NumberUnit SizePrice (USD)Quantity
EPX01A-12168-90196 tests
Price: 302.00
Your price:
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Human IL-20 Simplex ProcartaPlex Kit measures IL-20 protein and is designed to be combinable with other Simplex kits so that you can create your own multiplex panel that utilizes Luminex xMAP technology for protein detection/quantitation. When combining multiple Simplex kits (i.e., when you are not using a pre-configured Multiplex Panel), only one buffer kit (sold separately) is needed for each assay plate regardless of plex size.

About ProcartaPlex Assays for the Luminex Platform

ProcartaPlex immunoassays are based on the principles of a sandwich ELISA, using two highly specific antibodies binding to different epitopes of one protein to quantitate all protein targets simultaneously using a Luminex instrument. ProcartaPlex multiplex assays require as little as 25 μL of plasma or serum, or 50 μL of cell culture supernatant, and just four hours to obtain analyzed results.

More results per sample—measure multiple protein targets simultaneously in a single 25–50 μL sample

Well-established Luminex technology—highly referenced multiplexing platform for protein detection and quantitation

ProcartaPlex assays utilize Luminex xMAP (multianalyte profiling) technology for the simultaneous detection and quantitation of up to 80 protein targets in a single 25–50 μL sample from plasma, serum, cell culture supernatants, and other bodily fluids.

The Luminex beads in the ProcartaPlex assay are internally dyed with precise proportions of red and infrared fluorophores to create spectrally unique signatures that can be identified by the Luminex xMAP detection systems (e.g. Luminex 200, FLEXMAP 3D, and MAGPIX). Similar to a sandwich ELISA, the ProcartaPlex assay uses matched antibody pairs to identify the protein of interest. In a multiplexed assay, each spectrally unique bead is labeled with antibodies specific for a single target protein, and bound proteins are identified with biotinylated antibodies and streptavidin–R-phycoerythrin (RPE). The conjugation of protein-specific antibodies to a distinct bead allows for analysis of multiple targets in a single well.

The most significant difference between a ProcartaPlex assay and ELISA is that the capture antibody in the ProcartaPlex assay is conjugated to a bead and not adsorbed to the microplate well, so the ProcartaPlex assay reagents are free-floating in the solution. For detection, the Luminex 200 instrument, for example, contains two lasers, one to distinguish the spectral signature of each bead and the second to quantify the amount of RPE fluorescence, which is proportional to the amount of protein present in the sample. ProcartaPlex multiplex assays can profile more target proteins using significantly less sample in the same time that it takes to perform a traditional sandwich ELISA.

ProcartaPlex multiplex panels are available in multiple formats across six species (human, mouse, rat, nonhuman primate, porcine, and canine). Visit thermofisher.com/procartaplex for more information, including a comprehensive list of individual protein targets.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Type
Simplex Kit
Quantity
96 Tests
Species
Human
Combinability
Combinable
Bead Type
IL-20 [26]
Assay Sensitivity
See Certificate of Analysis
Assay Range
As determined for Lot 1: 12.26 - 50,200 pg/mL
Format
Simplex Kit
For Use With (Equipment)
Luminex™ Instruments
Gene
interleukin 20
Gene Alias
IL-20, IL10D, ZCYTO10
Gene ID (Entrez)
50604
Gene Symbol
IL20
Product Line
ProcartaPlex™
Protein
Interleukin-20
Protein Subtype
IL-20, Cytokine Zcyto10
Sample Type
Serum, Plasma, Cell Culture Supernatants
Sample Volume
Serum, Plasma: 25 μL; CCS: 50 μL
UniProt ID
Q9NYY1
Shipping Condition
Wet Ice
Research Area
Immunology

Contents & Storage

• 1 vial Capture Beads (50X)
• 1 vial biotinylated Detection Antibody (50X)
• 2 vials Human Standard Mix E (lyophilized)

Store at 2°C to 8°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text