ProcartaPlex™ NHP Control Set, 37plex
Invitrogen™

ProcartaPlex™ NHP Control Set, 37plex

The ProcartaPlex Non-Human Primate Control Set 37plex contains a high and low control for all analytes included in the Non-Human Primate Cytokine/Chemokine/Growth Factor 37-Plex ProcartaPlex Panel.
Have Questions?
Catalog NumberQuantity
EPX370-40045-CTR1 set
Catalog number EPX370-40045-CTR
Price (EUR)
252,00
Each
Quantity:
1 set
Price (EUR)
252,00
Each

The ProcartaPlex Non-Human Primate Control Set 37plex contains a high and low control for all analytes included in the Non-Human Primate Cytokine/Chemokine/Growth Factor 37-Plex ProcartaPlex Panel. For a list of analytes included in this set, please refer to the TDS in the Documents section.

Reported application

  • Multiplex Immunoassay
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)Luminex™ Instruments
Product LineProcartaPlex
Shipping ConditionWet Ice
Product TypeMultiplex Control Set
Quantity1 set
SpeciesNon-Human Primate
Unit SizeEach
Contents & Storage

• 1 vial NHP Control High A
• 1 vial NHP Control High B
• 1 vial NHP Control Low A
• 1 vial NHP Control Low B

Store at 2°C to 8°C.