Invitrogen™

MCP-2 Human ProcartaPlex™ Simplex Kit, Platinum

Catalog number:  EPXP010-12169-901

 Related applications:

Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Platinum ProcartaPlex™ Immunoassays are our are highest validated bead based Multiplexing assay system for quantificaton of multiple protein biomarkers in a single sample.
Newly developed matrix type specific diluents for plasma and serum enable to overcome matrix effects and assure high performance specifications (e.g spike and dilution recovery results between 70-130%).
Our Platinum ProcartaPlex assays use Luminex™ xMAP technology for the multi-analyte detection of secreted proteins including cytokines, chemokines, growth factors and other proteins. Platinum ProcartaPlex™ assays are subject to the same stringent and robust development and manufacturing as are our extensively validated Platinum and Instant ELISA assays.

Platinum ProcartaPlex Simplex kits can only be combined with one another.

Reactivity/Species
Human

Suitable Sample Types
Serum, Plasma

Sample Volume
Serum, Plasma: 25 µL

Reported Application
Multiplex Immunoassay

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Assay Range: As determined for Lot 1: 0.56-2300 pg/mL
Assay Sensitivity: See product documentation
Bead Region: 37
Former Identifiers: eBioscience
Gene Name: C-C motif chemokine ligand 8
Gene Symbol: CCL8
Immunoassay Kit Format: Simplex Kit
Immunoassay Kit Grade: Platinum
Protein Name: C-C motif chemokine 8
Sample Volume: Serum, Plasma: 25 µL
Target (Kit Named): MCP-2
Target List: MCP-2 (CCL8)
Target Species (Named): Human
Target Species (Validated): Human
UniProt ID: P80075
Detection Method: Fluorescent
For Use With (Equipment): Luminex™ Instruments
Gene Aliases: HC14, MCP-2, MCP2, SCYA10, SCYA8
Gene ID: 6355
Label or Dye: R-PE
Product Line: ProcartaPlex™
Product Size: 96 tests
Sample Type (Specific): Plasma, Serum
System: Luminex™

Contents & storage

1 vial Antibody Magnetic Beads (50X) coated with monoclonal antibody to human MCP-2 (CCL8)
1 vial Biotin-Conjugate (50x) anti-human MCP-2 (CCL8) monoclonal antibody

Store at 2-8°C.

Documents

Manuals & protocols