TheraPure™ Eam1104I, 3000 U, 150 μL
Thermo Scientific™

TheraPure™ Eam1104I, 3000 U, 150 μL

Thermo Scientific TheraPure in vitro transcription (IVT) enzymes are designed for advanced research and early development of mRNA therapeutics and vaccines.
Have Questions?
Catalog number ER023T1
Price (USD)
318.00
Each
Price (USD)
318.00
Each
Thermo Scientific TheraPure in vitro transcription (IVT) enzymes are designed for advanced research and early development of mRNA therapeutics and vaccines. These products offer full workflow solutions from template preparation to RNA synthesis and are recommended for process research and optimization, proof of concept, and preclinical studies.

The high purity of TheraPure IVT enzymes is ensured by precise quality testing that includes confirmation that that the enzymes are nuclease- and endotoxin-free. In addition, the manufacturing process of TheraPure products follows ISO9001 and ISO13485 standards.

Features

  • Convenient stand-alone enzymes for easy workflow optimization
  • Animal-origin-free (AOF) raw materials used in manufacturing
  • No risk of prion or animal virus contamination
  • Reproducible and consistent results with validated quality testing
  • Regulatory compliant—β-lactam-free manufacturing technology
  • Suitable for translational research
  • Facilitates transition to GMP during mRNA vaccine development process
  • Scalable and available as OEM product

The exceptional quality of TheraPure products is supported by:

  • Certificates of analysis
  • Certificates of origin
  • TSE/BSE statement
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Compatible Buffer10X Tango Buffer
Cut SiteCTCTTC(1/4)^
For Use With (Application)In vitro Transcription
Inactivation Temperature65°C
Incubator Temperature37°C
Product TypeRestriction Enzyme
Quantity3000 U, 150 μL
Concentration20 U/μL
EnzymeEam1104I
FormLiquid
Methylation SensitivityNot Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive, Not CpG Methylation-Sensitive
Optimal Reaction Temperature37°C
Product LineTheraPure
Research CategoryDrug Development
Sensitive to Heat InactivationYes
Type IIS REYes
Unit SizeEach
Contents & Storage
Store at -20°C.