SgsI (AscI) (10 U/μL)
SgsI (AscI) (10 U/μL)
SgsI (AscI) (10 U/μL)
Thermo Scientific™

SgsI (AscI) (10 U/μL)

5' G G ↓C G C G C C 3'詳細を見る
製品番号(カタログ番号)数量
ER18921500 U
ER1891300 U
製品番号(カタログ番号) ER1892
価格(JPY)
39,700
キャンペーン価格
Ends: 26-Jun-2026
66,300
割引額 26,600 (40%)
Each
数量:
1500 U
一括またはカスタム形式をリクエストする
5'  G  G ↓C  G  C  G  C  C   3' 
3'  C  C  G  C  G  C ↑G  G   5' 

Thermo Scientific SgsI(AscI)制限酵素は、GG^CGCGCC部位を認識し、その部位で切断します。Tangoバッファー中で37℃の場合に最適な切断が起こります。酵素活性、二重消化の条件、およびこの制限酵素や他の制限酵素の熱不活性化の表については、制限酵素の反応条件を参照してください。注:迅速なDNA消化のためのFastDigest酵素としてもご利用いただけます。アイソシゾマー:AscI、PalAI。

Thermo Scientificの従来の制限エンドヌクレアーゼは、高品質な制限酵素の大規模なコレクションで、ファイブバッファーシステムのいずれかのバッファーで機能するように最適化されています。また、二重消化に適した汎用的なTangoバッファーも提供されています。すべての酵素は、推奨されるバッファーおよび反応条件で100%の活性を示します。一貫した性能を確保するため、Thermo Scientificの制限酵素反応バッファーには、BSAがあらかじめ混合されています。これにより、多くの酵素の安定性が向上し、DNA調製物に存在する汚染物質が結合されます。

特長

• 優れた品質 — 厳格な品質管理と業界をリードする製造プロセス
• 便利なカラーコードで色分けされたファイブバッファーシステム
• 二重消化用の汎用的なTangoバッファー付き
• BSAは反応バッファーに事前に混合済み
• 広範な制限エンドヌクレアーゼ特異性

アプリケーション

• 分子クローニング
• 制限部位マッピング
• ジェノタイピング
• サザンブロッティング
• 制限酵素断片長多型(RFLP)
• SNP

注記:メチル化感受性については、製品仕様を参照してください。
研究用途にのみご使用ください。診断目的には使用できません。
仕様
適合バッファー10XバッファーTango
製品タイプ制限酵素
数量1500 U
濃度10 U/μL
酵素SgsI(AscI)
メチル化感受性CpG Methylation-Sensitive, Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive
最適反応温度37℃
研究カテゴリー従来のクローニング
熱不活性化への感受性
IIS RE型不可
Unit SizeEach

よくあるご質問(FAQ)

Can I double digest my DNA using a Thermo Scientific conventional restriction enzyme and a FastDigest restriction enzyme?

For optimal results with fast reaction and 100% buffer compatibility, we highly recommend using FastDigest restriction enzymes in double digestion. In certain cases however, it may be possible to perform double digestion using a mix of Thermo Scientific conventional and Fastdigest restriction enzymes. For specific recommendations, please contact our technical service with detailed information about the enzymes and DNA template you plan to use.

Why do you recommend only 2 µL of 10X Reaction Buffer when digesting unpurified PCR product in a 30 µL reaction?

We recommend only 2 µl 10X Buffer in digestion of unpurified PCR products in 30 ul since salts and ions from the PCR reaction would be carried over to the digestion reaction.

Find additional tips, troubleshooting help, and resources within our Restriction Enzyme Cloning Support Center.

What are key factors promoting star activity?

Star activty may be contributed by:

• Prolonged incubation
• High enzyme concentration
• High glycerol concentration (usually 5% or higher)
• Small reaction volume

Find additional tips, troubleshooting help, and resources within our Restriction Enzyme Cloning Support Center.

Unexpected DNA bands were observed on agarose gel electrophoresis after restriction digestion. What may have caused this?

Unexpected cleavage patterns may be caused by the following reasons:

• Star activity of the restriction enzyme: Make sure to follow the reaction recommendations as specified in the protocol. Star activity may be improved by changing several key factors such as decreasing the reaction time, increasing the reaction volume, and decreasing the enzyme amount.

• Partial or incomplete cleavage (incomplete restriction reaction): Efficiency of the enzyme can be improved by adding more enzyme, prolonging the reaction time, or purifying DNA samples to remove inhibitory contaminants.

• Contamination with non-specific endonucleases: Non-specific endonucleases may be introduced to the DNA sample and/or the enzyme from improper handling, pipetting, etc.

•Improper reaction setup: Mix the digestion reaction thoroughly.

Find additional tips, troubleshooting help, and resources within ourRestriction Enzyme Cloning Support Center.

What are possible reasons for incomplete/failed restriction digestion?

The main reason for DNA cleavage reaction failure is the presence of contaminating inhibitors in the template DNA (for example: phenol, chloroform, detergents, ethanol, excess salts, EDTA, etc.). The best way to troubleshoot is to perform control reactions:

1) negative control (experimental DNA in the reaction buffer without the restriction enzyme) to access degradation of DNA by contaminants in the DNA template and/or reaction buffer
2) positive control reaction I (digestion of highly pure control DNA with the restriction enzyme) to access reaction conditions and enzyme activity
3) positive control reaction II (highly pure control DNA + experimental DNA + Restriction Enzyme) to access possible issues with the experimental DNA.

In addition, please check for sensitivity of the restriction enzymes to template DNA methylation.

Find additional tips, troubleshooting help, and resources within our Restriction Enzyme Cloning Support Center.