Thermo Scientific™
Hydrophobic Printed Well Slides, 12-well, 5mm
 Related applications:
Anatomical Pathology
Virtually eliminate cross-contamination by keeping fluids in place with the hydrophobic coating on Thermo Scientific™ Hydrophobic Printed Well Slides. These hydrophobic slides have a durable printed surface that is inert to common laboratory chemicals and reagents.
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Back to top
Description
  • Ideal for use in PCR applications
  • Autoclavable and can withstand PCR and sterilization processes
Documents
Manuals & protocols