ER3 EPD Reader
ER3 EPD Reader
ER3 EPD Reader
Thermo Scientific™

ER3 EPD Reader

Read and store critical radiation dose information with the Thermo Scientific™ ER3 EPD Reader for the Thermo Scientific™ EPD TruDose Electronic Personal Dosimeter.
Have Questions?
Catalog NumberDescription
ER3EPDER3 EPD Reader
Catalog number ER3EPD
Price (USD)
-
Description:
ER3 EPD Reader
Read and store critical dose information in any environment where radiation exposure may be an issue, including medical, nuclear, research, and other health physic applications, with the Thermo Scientific™ ER3 EPD Reader. The ER3 EPD reader works with the Thermo Scientific™ EPD TruDose Electronic Personal Dosimeter, the EPD MK2+ dosimeter, and EPD N2 dosimeters and provides an interface between the EPD, user and an access control system. The ER3 EPD reader is available as a wall mounted version or as an exit reader mounted inside a personnel contamination monitor.
  • Infra-red optical link to EPD TruDose Electronic Personal Dosimeter, operates automatically when an EPD is presented to the optical port
  • LCD visual display
  • Supports temporary network offline mode by buffering data
  • Microprocessor based Audible beeper
  • Optional interface control port used for door or turnstile control
  • Serial interface to personnel ID card reader
  • Numeric keypad to enable data such as personnel ID or work permit numbers to be entered
  • Ethernet port for interfacing to site network
  • Messages displayed in either English or Chinese
Specifications
Certifications/ComplianceER3
Depth (Metric)75 mm, maximum from wall surface
DescriptionER3 EPD Reader
EMC StandardsEN61326-1:2013
Height (Metric)261 mm (Including Cable Cover)
Humidity10% to 85% at 29°C
Mains FuseT1A 250 V, 1 A TIME DELAY, 5x20 mm
Max. Power20 W
Operating Temperature (English)32°F to 122°F
Operating Temperature (Metric)0°C to 50°C
Power Supply100/240 VAC and 50/60 Hz
SafetyEN61010-1:2010 equipment is for indoor use only
SoftwareER-3 connects via the network to the Dose Management and Control Software (webREMS). Can be configured locally at the ER3.
Storage Temperature (English)-4°F to +122°F
Storage Temperature (Metric)-20°C to +70°C
Weight (English)8.81 lb.
Weight (Metric)4 kg
Width (English)10.86 in.
Width (Metric)276 mm
For Use With (Equipment)EPD MK2+, EPD N2, EPD TruDose (BG and G types)
Unit SizeEach