TNF alpha Rat ELISA Kit

Catalog number: ER3TNFA2

Invitrogen™  Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Assay TNFα (rat tumor necrosis factor alpha) levels in rat serum and culture supernatant samples with our Thermo Scientific Rat TNF alpha ELISA Kit.

Features of the Rat TNF alpha ELISA Kit:

Target: natural and recombinant rat TNFα (rat tumor necrosis factor alpha)
Format: colorimetric ELISA (TMB substrate) in clear 96-well strip plates
Measurement: absorbance at 450nm minus absorbance at 550nm
Assay Range: 31 to 2500 pg/mL
Sensitivity: < 15 pg/mL
Precision: intra-assay CV < 10%; inter-assay CV < 10%
Sample Types: cell culture supernatant, serum, plasma (EDTA, sodium citrate and heparin)
Total Assay Time: 3 hours
Sample Size: 50 µL
Specificity: no significant cross-reaction with human TNFα, human TNFβ or porcine TNFα. Mouse TNFα cross-reacts weakly (< 3%).

The Rat TNF alpha ELISA Kit uses pre-titered, matched pairs of coating and detection antibodies to achieve sensitive and accurate measurement of tumor necrosis factor alpha (TNF-alpha, TNFα) in samples at 50 µL per well. Also included are calibrated recombinant protein standard, streptavidin-HRP, TMB substrate solution and stop solution. The complete kit contains precoated 96-well strip plates and lot-optimized reagents. All components undergo quality control testing to minimize lot-to-lot variability.

Includes:
Complete kit contains pre-coated antibody-plates, detection antibodies, buffers, diluents, standards, and substrates.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Assay Sensitivity: See product documentation
Detection Method: Colorimetric
For Use With (Equipment): Microplate Reader
Former Identifiers: Pierce
Gene Aliases: RATTNF, TNF-alphaa, Tnf
Gene ID: 24835
Gene Name: tumor necrosis factor
Gene Symbol: TNF
Immunoassay Kit Format: Coated ELISA Kit
Label or Dye: HRP (Horseradish Peroxidase)
Product Size: 2 x 96 tests
Protein Name: tumor necrosis factor
Target (Kit Named): TNF alpha
Target Species (Named): Rat
Target Species (Validated): Rat
UniProt ID: P16599

Contents & storage

Pre-coated plate(s), standard or calibrator, detector antibody, HRP conjugate, diluents, wash buffer, substrate(s), and stop solution. Store kit at 2-8°C. See product manual for detailed contents and storage conditions for maximum stability.

Documents

Manuals & protocols