Thermo Scientific™
EASY-Spray™ HPLC Columns
 Related applications:
Industrial Chromatography
Ensure robust nano and capillary flow LC-MS with Thermo Scientific™ EASY-Spray™ LC Columns. The integrated column/emitter design eliminates dead volume and is temperature-controlled for maximum reliability and performance. Rigourously tested to ensure maximum quality, these columns are designed for maximum simplicity and ease-of-use.
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Back to top
Description
The unique EASY-Spray design provides uncompromised performance and ease-of-use in nano and capillary LC-MS proteomics applications. Features include the following:
  • Precision positioned glass emitter
  • Integrated precision designed union
  • Integrated temperature control
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Documents
Manuals & protocols