ELISA Systems™ Egg Residue Detection Kit for Food Allergen Testing
ELISA Systems™ Egg Residue Detection Kit for Food Allergen Testing
ELISA Systems™ Egg Residue Detection Kit for Food Allergen Testing
ELISA Systems™ Egg Residue Detection Kit for Food Allergen Testing
Thermo Scientific™

ELISA Systems™ Egg Residue Detection Kit for Food Allergen Testing

Rapidly detect and quantify egg material in uncooked food and wine.
Have Questions?
카탈로그 번호수량
ESERD-4848 Tests
카탈로그 번호 ESERD-48
제품 가격(KRW)
-
수량:
48 Tests
Simplify allergen testing with ELISA Systems™ Egg Residue Detection Kit which helps ensure compliance with food product labeling requirements. The enzyme-linked immunosorbent assay is designed to deliver simple, sensitive, and fast detection and quantification of casein as an indicator of the presence of milk or milk products. This complete, ready-to-use kit is developed and optimized for rapid and reliable test results that significantly reduce the time required to screen appropriate food products for the presence of egg residue.
Hen’s egg is one of the more frequent causes of food hypersensitivity in infants and young children.1, 2

Use ELISA SystemsEgg Residue Detection Kit for the rapid detection and quantification of egg residue in food and environmental samples. The kit is primarily suitable for the detection of residues of uncooked egg and is also suitable for wine testing.

  • Fast: Sample preparation to result in under 60 minutes for rapid test turnaround and actions
  • Simple: Ready-to-use liquid reagents and concentrates, plus common handling procedure for all kits, make ELISA Systems Assays easy to implement and run
  • Robust: Ready-to-use liquid controls and allergen-free non-control reagents mean reduced risk of cross contamination that could lead to false-positive results
  • Flexible: Use for qualitative or quantitative testing—a single kit for more applications
  • Sensitive: Detect proteins or protein residues, including heat stable proteins, at low levels for improved assurance of product safety
  • Supported: Our experienced technical team is ready to assist you and help mitigate delays in test reporting


Two of the main allergens in egg white are Ovalbumin (54%) and Ovomucoid (11%)3. Ovalbumin is the major allergen present in the egg white. Ovomucoid is allergenic in minute quantities and is not coaguable by heat4.

Ovalbumin (because of its allergenic properties) and Ovomucoid (because of its allergenic and heat stable properties) were chosen as the Egg protein indicators for the ELISA Systems Egg Residue Detection Kit.

All results should be interpreted as part of a HACCP plan for food allergens. Any sample returning a positive result should be regarded as a presumptive result, and confirmation or further testing should be performed.

Not all products are available for sale in all territories. Please inquire.

ELISA Systems™ Food Allergen Test Kits are manufactured by ELISA Systems Pty Ltd.

ELISA Systems is a trademark of ELISA Systems Pty Ltd.

General References:

  1. Sampson. H.A., McCakill. C.C. Food hypersensitivity and atopic dermatitis: evaluation of 113 patients. J Pediatr 1985; 107:669-75.
  2. Bock. S.A., Atkins.F.M. Patterns of food hypersensitivity during sixteen years of double-blind, placebo controlled food challenges. J Pediatr 1990;117:561-7.
  3. Bernhisel-Broadbent J, Dintzis. H.M., Dintzis. R.Z., Sampson.H.A.. Allergenicity and antigenicity of chicken egg ovomucoid (Gal d III) compared with ovalbumin (Gal d I) in children with egg allergy and in mice. J Allergy Clin Immunol 1994;93: 1047-59.
  4. Atsuo Urisu, Hitoshi Ando, Yutaka Morita, Eiko Wada, Takehiko Yasaki, Kazue Yamada, Katsuhide Komada, Shimpei Torii, Masahiro Goto, Toshio Wakamatsu. Allergenic activity of heated and ovomucoiddepleted egg white. J Allergy Clin Immunol 1997;100: 171-6.
사양
설명ELISA Systems Egg Residue Detection Kit
검출 가능한 분석물질Ovalbumin and Ovomucoid Egg Protein
키트 구성품20X Extraction solution concentrate, 20X wash buffer solution concentrate, Enzyme conjugate, Negative Standard, Positive Standards, Stop solution, Substrate, Test strips
제품 유형Egg Residue Detection Kit
수량48 Tests
보관 요구 사항2°C to 8°C
이뮤노어세이 키트 형식ELISA (Standard)
Unit SizeEach