Heratherm Stability Chambers
Heratherm Stability Chambers
Heratherm Stability Chambers
Heratherm Stability Chambers
Heratherm Stability Chambers
Heratherm Stability Chambers
Heratherm Stability Chambers
Heratherm Stability Chambers
Thermo Scientific™

Heratherm Stability Chambers

Gain confidence in your product when testing stability and shelf life with robust Heratherm Stability chambers designed to meet a variety of quality control demands.
製品番号(カタログ番号)ドアスタイル容量(メートル法)
ESS250Solid outer door with inner glass door247 L
ESS750Solid (outer), Glass (inner)750 L
ESG750Glass750 L
ESG250Glass door247 L
製品番号(カタログ番号) ESS250
価格(JPY)
-
ドアスタイル:
Solid outer door with inner glass door
容量(メートル法):
247 L
Temperature and humidity controls, well suited for stability testing, shelf life testing, and environmental testing.
  • Broad relative humidity range to cover many applications, including humidity levels below ambient.
  • Environmentally friendly water-efficient humidification system.
  • Minimal condensate in standard applications.
  • Integrated water tank; fixed water connection for in-house purified water option available.
  • Energy-efficient Peltier technology supports temperature stability and uniformity with lower energy usage compared to traditional compressors and without hazardous refrigerant.
  • Two access ports for independent data capture, and internal documentation of set and actual parameter values, along with event log to meet 21 CFR Part 11 requirements for process documentation.
  • Data retrieval via USB stick, Ethernet connection, or Thermo Cloud via OPC-UA standard.
  • Sample and process protection: Assign personalized access with user IDs; restrict access with standard door lock, or optional automated door lock; various alarm options to prevent operator of process issues.
  • Intuitive touchscreen interface with easy programmability of temperature cycles and process duration.
  • Simplified setup with no technician required; install using standard power cord.
  • IQ/OQ qualifications and other services available; visit thermofisher.com/chamberservice.
  • Inner glass door keeps environment stable, even when opening outer door. Glass door also available.
  • Rounded corners and removable shelving offer easy cleaning to help prevent cross-contamination.
  • Duplex interior electrical sockets for use of stirrers, shakers or other equipment inside the chamber are available as a factory-installed option. Choose from a variety of country-specific sockets. (Ordered with separate order code)
仕様
アラームタイプAudible and Visible
容量(ヤードポンド法)8.8 cu.ft.
容量(ヤードポンド法)棚66.1 lb.
容量(メートル法)棚30 kg
認証/コンプライアンスUL, CE
データ出力USB, Thermo Cloud via OPC UA
奥行き(ヤードポンド法)31.1 in.
奥行き(ヤードポンド法)内部20.4 in.
奥行き(メートル法)790 mm
奥行き(メートル法)内部519 mm
ディスプレイ7 in. Touchscreen user interface
ドアスタイルSolid outer door with inner glass door
電気的要件100/240V, 50/60Hz
使用対象(アプリケーション)Stability testing, environmental testing, storage
高さ(ヤードポンド法)44.4 in.
高さ(ヤードポンド法)内部31.5 in
高さ(メートル法)1128 mm
高さ(メートル法)内部800 mm
湿度範囲10 to 90% RH
湿度発生源Removable water tank
照明No
キャスター数None
プラグタイプNema 5-20P
ポート2 access ports
プログラム可能Temperature and humidity ramping
棚面積(ヤードポンド法) 3.05 ft.2
棚面積(メートル法)0.28 m2
棚材質Stainless Steel (304/1.4301)
出荷重量(ヤードポンド法)459 lb.
出荷重量(メートル法)208 kg
温度制御Microprocessor - programmable
温度分解能0.1°C
温度安定性≤ ±0.1°C @ 20°C to 37°C
温度均一性≤ ±0.5°C @ 20°C to 37°C
タイプStability Chamber
保証24 months
水消費量≤ 0.25 L per day - Value depends on settings, usage (e.g. number of door openings), and environmental conditions
水比抵抗0.05 - 1 MΩ
ワット数900 W
幅(ヤードポンド法)35.5 in.
幅(ヤードポンド法)内部26 in.
幅(ヤードポンド法)棚24.2 in.
幅(メートル法)901 mm
幅(メートル法)内部660 mm
幅(メートル法)棚615 mm
容量(メートル法)247 L
チャンバー材質Powder-coated Cold Rolled Steel
材料Powder-coated Cold Rolled Steel
棚数2
相対湿度10 to 90%
温度範囲(摂氏)0 to 70°C
電圧100/115/220/230 V
Unit SizeEach