Thermo Scientific™ QSP™ Macro Volume Pipette Tips
Actual product may vary
Thermo Scientific™ QSP™ Macro Volume Pipette Tips
Thermo Scientific™ QSP™ Macro Volume Pipette Tips
Thermo Scientific™ QSP™ Macro Volume Pipette Tips
Thermo Scientific™

Thermo Scientific™ QSP™ Macro Volume Pipette Tips

Thermo Scientific™ QSP™ Macro Volume Pipette Tips are ideal for transferring large volumes of liquid
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호부피(미터법)멸균오토클레이브 가능형식제품 사이즈
F090100S-5000-Q5000 μLSterileNot AutoclavableBulkCase of 1000
089-Q250 to 2500 μLNon-sterileAutoclavableCase of 2500
089-QPK
089-Q으로도 사용됨
250 to 2500 μLNon-sterileAutoclavablePack of 250
F090100S-5000-QPK
F090100S-5000-Q으로도 사용됨
5000 μLSterileNot AutoclavableBulkPack of 100
096-NXL-Q250 to 2500 μLSterileAutoclavableCase of 2500
096-NXL-QPK
096-NXL-Q으로도 사용됨
250 to 2500 μLSterileAutoclavablePack of 250
카탈로그 번호 F090100S-5000-Q
제품 가격(KRW)
781,000
온라인 행사
Ends: 30-Sep-2026
918,000
할인액 137,000 (15%)
Case of 1000
부피(미터법):
5000 μL
멸균:
Sterile
오토클레이브 가능:
Not Autoclavable
형식:
Bulk
Unit Size:
Case of 1000
제품 가격(KRW)
781,000
온라인 행사
Ends: 30-Sep-2026
918,000
할인액 137,000 (15%)
Case of 1000

Thermo Scientific™ QSP™ Macro Volume 250-2500 μL Pipette Tips are packaged in bulk bags and non-sterile clean claims are; Rnase/Dnase, DNA, pyrogen and endotoxin free. Specific fit for Rainin EDP2 pipette.

  • Sterile and non sterile 2.5ml, 5ml and 10ml tip volumes allow you to transfer bulk reagents with less effort
  • Universal fit and pipettor specific options meet broad pipette range
  • Filtered and Non-Filtered choices depending on application requirement
사양
클린 클레임Sterile Electron Beam free of: Rnase/Dnase, DNA, pyrogen, endotoxin, ATP, PCR Inhibitor and bioburden
오토클레이브 가능Not Autoclavable
필터링됨Filtered
형식Bulk
물질100% Virgin Polypropylene
멸균Sterile
팁 스타일Universal Fit
부피(미터법)5000 μL
Unit SizeCase of 1000