HyPerforma™ Process Development Automation Package, for 3 G3Lab Controller Systems
HyPerforma™ Process Development Automation Package, for 3 G3Lab Controller Systems
Thermo Scientific™
HyPerforma™ Process Development Automation Package, for 3 G3Lab Controller Systems
The Thermo Scientific HyPerforma Automation Packages are specifically designed for PD scale bioprocessing applications. The system features Thermo Scientific TruBioRead more
Have Questions?
Catalog number F100-2679-003
Price (KRW)
-
Request A Quote
The Thermo Scientific HyPerforma Automation Packages are specifically designed for PD scale bioprocessing applications. The system features Thermo Scientific TruBio software that facilitates easy, reliable, and repeatable process development cell culture processes.
HyPerforma PD Automation Package features include:
• Open architecture capabilities to integrate with vessels from other suppliers (Sartorius, Eppendorf, Millipore,and Applikon)
• The ability to build and manage complex, multi-feed dosing strategies that provide for third-party peripheral integration as needed
Applications
• Suspension and adherent cell culture processes
• Fermentation processes
• PD scale to full cGMP manufacturing workflows
Specifications
Operating SystemEmerson™ DeltaV™ system
Shipping ConditionRoom Temperature
Product LineHyPerforma™
Unit SizeEach
Contents & Storage
• TruLogic Base Cabinet (NEMA 12/IP54, stainless steel cabinet)
• DeltaV MQ Controller, M-Series, VE3008 (1)
• Serial card (2)
• TruBio Media Version 13, Release 5.0.X
• DeltaV Software Media Pack
• ProPlus 25 Device Signal Tag (DST)
• Analog Control Output 25 DST
• Guardian Support,1 year, 100 DST
• Microsoft Excel software, one license
• ProPlus Workstation (PowerEdge tower server, RAID 1, Windows Server 2016)
• Application Station (PowerEdge tower server, RAID 10, Windows Server 2016)
• 22 in. Wide-Screen Monitor (required for TB 5.0 batch) (2)
• Cable Assembly, Cat5e, Yellow Boot, TruLogic, PRI Network, 20 ft (6 m) (2)
• Cable Assembly, Cat5e, Black Boot, TruLogic, SEC Network, 20 ft (6 m) (2)
• Application/base station license (250 DeltaV for OPC-DA plus 250 DeltaV Historian Parameters) (required for Application Station use)
• Application Station/Historian license scale-up (1000 DeltaV Historian Parameters)
• DeltaV equipment crating/packaging
• Audit trail