HyPerforma™ G3Lab Controller, for HyPerforma Rocker and Single-Use Bioreactors (non-GMP)
Thermo Scientific™

HyPerforma™ G3Lab Controller, for HyPerforma Rocker and Single-Use Bioreactors (non-GMP)

The Thermo Scientific G3Lab Controller has been specifically designed for PD scale bioprocessing applications. It can control most brands ofMás información
Have Questions?
Número de catálogo F100-2694-002
Precio (EUR)
-
The Thermo Scientific G3Lab Controller has been specifically designed for PD scale bioprocessing applications. It can control most brands of single-use or autoclavable bioreactors or fermentors with a capacity of ≤50 L, including stirred-tank and rocking models. The controller operates using proprietary Thermo Scientific TruBio software, which provides easy process scale-up/scale-down and the configurability to scale your control strategy along with your process. The enclosure contains state-of-the-art transmitters along with power supplies, pumps, input/output (I/O) modules, and the hardware required to connect to the control network, providing maximum control capability.

G3Lab Controller features include:
• Open architecture capabilities to integrate with vessels from other suppliers (Sartorius, Eppendorf, Millipore, and Applikon)
• A common control platform throughout a product’s life cycle
• Flexible upstream TruBio software powered by the Emerson™ DeltaV™ system
• Nonproprietary, industry-standard, cGMP-validated (with validation package) distributed control system (DCS) that provides networked multiple-vessel control capability

Applications
• Suspension and adherent cell culture processes
• PD scale to full cGMP manufacturing workflows

Especificaciones
MaterialAluminum (Top Cover)
Certificaciones/conformidadTested and verified as meeting CE standards EN-60101 and EN-61326
Tensión110/240 VAC 50/60 Hz
Dimensiones438.4 x 240 x 482.6 mm (17 1/4 x 9 3/8 x 19 in.)
Sistema operativoEmerson™ DeltaV™ system
Condiciones de envíoRoom Temperature
Peso (métrico)16.42 kg/21.4 kg
Línea de productosHyPerforma™
Unit SizeEach
Contenido y almacenamiento
• HyPerforma G3Lab Controller
• TruBio license, with 4 MFCs
• 4 Watson-Marlow series 114 pumps
• Transmitter for pH+dO2 Sensor (optical pH and DO)
• TruSens transmitter for RTD, and all necessary cables; for HyPerforma Rocker Bioreactor only