HyPerforma™ G3Lab Controller, for HyPerforma Glass Bioreactor (GMP)
Thermo Scientific™
HyPerforma™ G3Lab Controller, for HyPerforma Glass Bioreactor (GMP)
The Thermo Scientific G3Lab Controller has been specifically designed for PD scale bioprocessing applications. It can control most brands ofRead more
Have Questions?
Catalog number F100-2695-001
Price (USD)
-
The Thermo Scientific G3Lab Controller has been specifically designed for PD scale bioprocessing applications. It can control most brands of single-use or autoclavable bioreactors or fermentors with a capacity of ≤50 L, including stirred-tank and rocking models. The controller operates using proprietary Thermo Scientific TruBio software, which provides easy process scale-up/scale-down and the configurability to scale your control strategy along with your process. The enclosure contains state-of-the-art transmitters along with power supplies, pumps, input/output (I/O) modules, and the hardware required to connect to the control network, providing maximum control capability.
G3Lab Controller features include:
• Open architecture capabilities to integrate with vessels from other suppliers (Sartorius, Eppendorf, Millipore, and Applikon)
• A common control platform throughout a product’s life cycle
• Flexible upstream TruBio software powered by the Emerson™ DeltaV™ system
• Nonproprietary, industry-standard, cGMP-validated (with validation package) distributed control system (DCS) that provides networked multiple-vessel control capability
Applications
• Suspension and adherent cell culture processes
• PD scale to full cGMP manufacturing workflows
Specifications
MaterialAluminum (Top Cover)
Voltage110/240 VAC 50/60 Hz
Dimensions438.4 x 240 x 482.6 mm (17 1/4 x 9 3/8 x 19 in.)
Operating SystemEmerson™ DeltaV™ system
Shipping ConditionRoom Temperature
Weight (Metric)16.42 kg/21.4 kg
Product LineHyPerforma™
Unit SizeEach
Contents & Storage
• HyPerforma G3Lab Controller
• TruBio license, with 4 MFCs
• 4 Watson-Marlow series 114 pumps
• TruSens transmitter for pH and DO, and all necessary cables; for HyPerforma Glass Bioreactor only; GMP