HyPerforma™ Commercial Automation Package, for 3 G3Lite Controller Systems
Thermo Scientific™
HyPerforma™ Commercial Automation Package, for 3 G3Lite Controller Systems
The Thermo Scientific HyPerforma GMP Automation Packages can be integrated with any single-use bioreactor such as the Thermo Scientific HyPerformaRead more
Have Questions?
Catalog number F100-2698-001
Price (CLP)
-
The Thermo Scientific HyPerforma GMP Automation Packages can be integrated with any single-use bioreactor such as the Thermo Scientific HyPerforma Single-Use Bioreactor (S.U.B.) (available in sizes ranging from 50 L to 2,000 L). The system features Thermo Scientific TruBio software that facilitates easy, reliable, and repeatable process development and commercial cell culture processes.
HyPerforma GMP Automation Package features include:
• Open architecture capabilities to integrate with vessels from other suppliers (Sartorius, Eppendorf, Millipore,and Applikon)
• The ability to build and manage complex, multi-feed dosing strategies that provide for third-party peripheral integration as needed
Applications
• Suspension and adherent cell culture processes
• Fermentation processes
• GMP manufacturing workflows
Specifications
Operating SystemEmerson™ DeltaV™ system
Shipping ConditionRoom Temperature
Product LineHyPerforma™
Unit SizeEach
Contents & Storage
• TruLogic Base Cabinet (NEMA 12/IP54, stainless steel cabinet)
• DeltaV MQ Controller, M-Series, VE3008
• Serial Card (2)
• TruBio Media Version 13, Release 5.0.X
• DeltaV Software Media Pack
• Microsoft Excel software, one license
• Small Form Factor Tower, Single Monitor, Windows 10 Enterprise loT 2016; Quad-Core CPU, 1 x 500 GB SATA Drive (monitor not included) (2)
• Base Station License
• Base Station Scale-Up License, 100 DST
• Process History View License
• TruBio Slave Dongle (or software key), Permanent (2)
• PlantWeb Experience 100 DST, v12.3 or Newer Software - one operator station/250pt OPC-DA on ProPlus/1 year foundation support included
• ProPlus Workstation (PowerEdge tower server, RAID 1, Windows Server 2016 for DV 13.3)
• Cable Assembly, cat5e, Yellow Boot, TruLogic, PRI Network, 20 ft (6 m) (3)
• Cable Assembly, cat5e, Black Boot, TruLogic, SEC Network, 20 ft (6 m) (3)
• 22 in. Wide-Screen Monitor (required for TB 5.0 batch) (4)
• Application/Base Station License (250 DeltaV for OPC-DA plus 250 DeltaV Historian Parameters)(required for Application Station use)
• Application Station/Historian License Scale-Up (1000 DeltaV Historian Parameters)
• Application Station (PowerEdge tower server, RAID 10, Windows Server 2016)
• Audit Trail