Product Image
Thermo Scientific™
DynaChrom™ Chromatography System Single-Use Fluid Transfer Assembly Set, 1/2 in.
DynaChrom Single-Use Fluid Transfer assemblies are designed to work with the DynaChrom Chromatography System. Available in 1/4-, 3/8-, 1/2-, andRead more
Have Questions?
Catalog NumberDiameter (English) Inner
F100-3600-0031/2 in.
Catalog number F100-3600-003
Price (KRW)
-
Diameter (English) Inner:
1/2 in.
DynaChrom Single-Use Fluid Transfer assemblies are designed to work with the DynaChrom Chromatography System. Available in 1/4-, 3/8-, 1/2-, and 3/4-inch sizes, they are optimized to match the flow rates with the single-use pump heads and single-use sensors. All wetted materials used in the flow path are verified for chemical compatibility and resistance to commonly used solvents/solutions.
Features of DynaChrom Single-Use Fluid Transfer assemblies include:
• BioTitan Retention Device enhances overall reliability and integrity by minimizing risk of leaks and failures at connection points
• In-line single-use sensors for flow and pressure measurements are provided at pump outlets
• Single-use flow cell with pH, conductivity, and temperature measurements provided pre-column and one with pH, conductivity, temperature, and UV measurements provided post-column. Multiple wavelength options available to suit chromatography process needs.
• Single-use valve manifold block flow paths matched to fluid transfer assemblies
• Four sub-assemblies in each fluid transfer assembly (depending on number of pumps). Sub-assemblies make unpacking and installation easier, are individually packaged, and can be ordered separately.
• Unique bubble trap design eliminates air bubbles in the system
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
Certifications/ComplianceGMP compliant, ADCF
For Use WithDynaChrom Chromatography System
For Use With (Application)Liquid Chromatography
GradeUSP Class VI
IncludesSensors, pumps, manifold blocks
Sterilization MethodIrradiation up to 40 kGy
Diameter (English) Inner1/2 in.
Diameter (English) Outer0.847 in.
Unit SizeEach