Phire Hot Start II DNA Polymerase, 200 reactions - FAQs
Phire Hot Start II DNA Polymerase, 200 reactions - FAQs
View additional product information for Phire Hot Start II DNA Polymerase - FAQs (F122L, F124S, F122S, F124L)
1 product FAQs found
Do Phire and Phusion Hot Start II DNA Polymerases need a separate activation step in the PCR protocol?
No separate activation step is required since Phire and Phusion Hot Start II DNA Polymerases are inactivated by a reversibly bound, specific Affibody ligand that dissociates during initial denaturation.