Search
View additional product information for Phire Hot Start II PCR Master Mixes - FAQs (F125S, F126S, F126L, F125L)
1 product FAQs found
No separate activation step is required since Phire and Phusion Hot Start II DNA Polymerases are inactivated by a reversibly bound, specific Affibody ligand that dissociates during initial denaturation.