Search
| 產品號碼 | Clean Claims | Volume (Metric) | Sterility | Unit Size |
|---|---|---|---|---|
| F129-96RS-1200Q | Sterile Electron Beam free of: Rnase/Dnase, DNA, pyrogen, endotoxin, ATP, PCR Inhibitor and bioburden | 200 μL | Sterile | Each |
| 115-96RN-Q | Non-Sterile free of: Rnase/Dnase, DNA, pyrogen and endotoxin | 200 μL | Non-sterile | Case of 4800 |
| 115-96RNS-Q | Sterile Electron Beam free of: Rnase/Dnase, DNA, pyrogen, endotoxin, ATP, PCR Inhibitor and bioburden | 100 μL | Sterile | Case of 4800 |
| 115-N-Q | Non-Sterile free of: Rnase/Dnase, DNA, pyrogen and endotoxin | 100 μL | Non-sterile | Case of 10000 |
| 116-N-Q | Non-Sterile free of: Rnase/Dnase, DNA, pyrogen and endotoxin | 1000 μL | Non-sterile | Case of 10000 |
| 116-R100N-Q | Non-Sterile free of: Rnase/Dnase, DNA, pyrogen and endotoxin | 1000 μL | Non-sterile | Case of 5000 |
| 116-R100NS-Q | Sterile Electron Beam free of: Rnase/Dnase, DNA, pyrogen, endotoxin, ATP, PCR Inhibitor and bioburden | 1000 μL | Sterile | Case of 5000 |
| 117-96RN-Q | Non-Sterile free of: Rnase/Dnase, DNA, pyrogen and endotoxin | 250 μL | Non-sterile | Case of 4800 |
| 117-N-Q | Non-Sterile free of: Rnase/Dnase, DNA, pyrogen and endotoxin | 250 μL | Non-sterile | Case of 10000 |
| 117-96RNS-Q | Sterile Electron Beam free of: Rnase/Dnase, DNA, pyrogen, endotoxin, ATP, PCR Inhibitor and bioburden | 250 μL | Sterile | Case of 4800 |
| F115-96RNS100-Q | Sterile Electron Beam free of: Rnase/Dnase, DNA, pyrogen, endotoxin, ATP, PCR Inhibitor and bioburden | 100 μL | Sterile | Each |
| F115-R100S200-Q | Sterile Electron Beam free of: Rnase/Dnase, DNA, pyrogen, endotoxin, ATP, PCR Inhibitor and bioburden | 200 μL | Sterile | Each |
| F116R100S1000-Q | Sterile Electron Beam free of: Rnase/Dnase, DNA, pyrogen, endotoxin, ATP, PCR Inhibitor and bioburden | 1000 μL | Sterile | Each |
| F11796RNS-200-Q | Sterile Electron Beam free of: Rnase/Dnase, DNA, pyrogen, endotoxin, ATP, PCR Inhibitor and bioburden | 200 μL | Sterile | Each |
| F11796RNS-100-Q | Sterile Electron Beam free of: Rnase/Dnase, DNA, pyrogen, endotoxin, ATP, PCR Inhibitor and bioburden | 100 μL | Sterile | Each |
| F117-96RS-300-Q | Sterile Electron Beam free of: Rnase/Dnase, DNA, pyrogen, endotoxin, ATP, PCR Inhibitor and bioburden | 300 μL | Sterile | Each |
| 115-R-Q | RNase/DNase, DNA, pyrogen and endotoxin free | 200 μL | Non-sterile | Case of 10 |
| 116-Q | RNase/DNase, DNA, pyrogen and endotoxin free | 200 μL | Non-sterile | Case of 10000 |
| 116-QPK 亦稱為 116-Q | RNase/DNase, DNA, pyrogen and endotoxin free | 200 μL | Non-sterile | Pack of 1000 |
| 116-R100-Q | RNase/DNase, DNA, pyrogen and endotoxin free | 200 μL | Non-sterile | Case of 5000 |
| 116-R100-QPK 亦稱為 116-R100-Q | RNase/DNase, DNA, pyrogen and endotoxin free | 200 μL | Non-sterile | Pack of 1000 |
| 116-R100S-Q | RNase/DNase, DNA, pyrogen and endotoxin, ATP, PCR Inhibitor and bioburden free | 200 μL | Sterile | Case of 5000 |
| 116-R100S-QPK 亦稱為 116-R100S-Q | RNase/DNase, DNA, pyrogen and endotoxin, ATP, PCR Inhibitor and bioburden free | 200 μL | Sterile | Pack of 1000 |
| T114R-Q | RNase/DNase, DNA, pyrogen and endotoxin free | 20 μL | Non-sterile | Case of 4800 |
| T114R-QPK 亦稱為 T114R-Q | RNase/DNase, DNA, pyrogen and endotoxin free | 20 μL | Non-sterile | Pack of 960 |
| T114RS-Q | RNase/DNase, DNA, pyrogen and endotoxin, ATP, PCR Inhibitor and bioburden free | 20 μL | Sterile | Case of 4800 |
| T114RS-QPK 亦稱為 T114RS-Q | RNase/DNase, DNA, pyrogen and endotoxin, ATP, PCR Inhibitor and bioburden free | 20 μL | Sterile | Pack of 960 |
| TF114-10-Q | RNase/DNase, DNA, pyrogen and endotoxin free | 200 μL | Non-sterile | Case of 4800 |
| TF114-20-Q | RNase/DNase, DNA, pyrogen and endotoxin free | 200 μL | Non-sterile | Case of 4800 |
| TF114-20-QPK 亦稱為 TF114-20-Q | RNase/DNase, DNA, pyrogen and endotoxin free | 200 μL | Non-sterile | Pack of 960 |
Thermo Scientific™ QSP™ Pipettor Specific Pipette Tips 200 μL natural, beveled pipette tips. Packaged in racks and non-sterile clean claims are; Rnase/Dnase, DNA, pyrogen and endotoxin free. Fits on Eppendorf pipettes.