Tissue Puncher - FAQs
View additional product information for Tissue Puncher - FAQs (F200S)
1 product FAQs found
What is your recommendation to take a sample from plant or animal tissue for Direct PCR?
In order to obtain best results from Direct PCR, it is important to use a very small amount of starting materials. The recommended size is 0.3-0.5 mm in diameter. We recommend using Tissue Puncher 0.3 mm (Cat. No. F-200S), which is a convenient tool for sampling and is suitable for both plant and animal tissues.