Thermo Scientific™

Target2™ Polypropylene Syringe Filters

Catalog number: F2513-9
Thermo Scientific™

Target2™ Polypropylene Syringe Filters

Catalog number: F2513-9
Adding a syringe filtration step prior to injection not only helps to ensure more consistent and reliable results; it also helps protect delicate instruments and prolongs column life. Available in sizes and membrane types to meet any application need, Thermo Scientific™ Target2™ Syringe Filters provide cleaner sample extracts by removing interfering materials and fine particles. Target2 Polypropylene Syringe Filters are a good choice for filtration of biological samples, or for filtration of aggressive organic solutions.
 
Catalog Number
F2513-9
Unit Size
Pack of 100
Diameter (Metric)
17 mm
Pore Size
0.45 μm
Type
Syringe Filter
Price (USD)
343.35
Your Price:
Availability
***
Full specifications
Quantity
Certifications/ComplianceTarget Polypropylene Syringe Filter Housings Meet The Requirements of 21 CFR 177.1520.
ColorWhite
DescriptionTarget2 Polypropylene Syringe Filters 0.45 µm, 17 mm, 100 Pk.
DisposableYes
MaterialPolypropylene
Material (Membrane)Polypropylene
Pore Size0.45 μm
SterilityNon-sterile
For Use With (Application)HPLC, SPE
Hold up Volume (After Air Purge)<35 μL
Housing MaterialMedical Grade, Solvent-Resistant, Virgin Polypropylene
PrefilterNo
Pressure Rating115 psi
Product LineTarget2
TypeSyringe Filter
Volume (Metric)<29 μL (Hold Up)
Diameter (Metric)17 mm
Diameter (Metric) Membrane17 mm
Unit SizePack of 100
Catalog Number
Unit Size
Pack of 100
Diameter (Metric)
30 mm
Pore Size
0.2 μm
Type
Syringe Filter
Price (USD)
397.95
Your Price:
Availability
***
Full specifications
Quantity
Certifications/ComplianceTarget Polypropylene Syringe Filter Housings Meet The Requirements of 21 CFR 177.1520.
ColorRoyal Blue
DescriptionTarget2 Polypropylene Syringe Filters 0.2 µm, 30 mm, 100 Pk.
DisposableYes
MaterialPolypropylene
Material (Membrane)Polypropylene
Pore Size0.2 μm
SterilityNon-sterile
For Use With (Application)HPLC, SPE
Hold up Volume (After Air Purge)<157 μL
Housing MaterialMedical Grade, Solvent-Resistant, Virgin Polypropylene
PrefilterNo
Pressure Rating150 psi
Product LineTarget2
TypeSyringe Filter
Volume (Metric)<137 μL (Hold Up)
Diameter (Metric)30 mm
Diameter (Metric) Membrane30 mm
Unit SizePack of 100
Catalog Number
Unit Size
Pack of 100
Diameter (Metric)
30 mm
Pore Size
0.45 μm
Type
Syringe Filter
Price (USD)
447.30
Your Price:
Availability
***
Full specifications
Quantity
Certifications/ComplianceTarget Polypropylene Syringe Filter Housings Meet The Requirements of 21 CFR 177.1520.
ColorWhite
DescriptionTarget2 Polypropylene Syringe Filters 0.45 µm, 30 mm, 100 Pk.
DisposableYes
MaterialPolypropylene
Material (Membrane)Polypropylene
Pore Size0.45 μm
SterilityNon-sterile
For Use With (Application)HPLC, SPE
Hold up Volume (After Air Purge)<157 μL
Housing MaterialMedical Grade, Solvent-Resistant, Virgin Polypropylene
PrefilterNo
Pressure Rating150 psi
Product LineTarget2
TypeSyringe Filter
Volume (Metric)<137 μL (Hold Up)
Diameter (Metric)30 mm
Diameter (Metric) Membrane30 mm
Unit SizePack of 100
Catalog Number
Unit Size
Pack of 100
Diameter (Metric)
30 mm
Pore Size
0.45 μm
Type
Syringe Filter with Pre-filter
Price (USD)
383.25
Your Price:
Availability
***
Full specifications
Quantity
Certifications/ComplianceTarget Polypropylene Syringe Filter Housings Meet The Requirements of 21 CFR 177.1520.
ColorWhite
DescriptionTarget2 Polypropylene Syringe Filters 0.2 µm, 30 mm, with Pre-Filter, 100 Pk.
DisposableYes
MaterialPolypropylene
Material (Membrane)Polypropylene
Pore Size0.45 μm
SterilityNon-sterile
For Use With (Application)HPLC, SPE
Hold up Volume (After Air Purge)<157 μL
Housing MaterialMedical Grade, Solvent-Resistant, Virgin Polypropylene
PrefilterYes
Pressure Rating150 psi
Product LineTarget2
TypeSyringe Filter with Pre-filter
Volume (Metric)<137 μL (Hold Up)
Diameter (Metric)30 mm
Diameter (Metric) Membrane30 mm
Unit SizePack of 100
Catalog Number
Unit Size
Pack of 100
Diameter (Metric)
4 mm
Pore Size
0.2 μm
Type
Syringe Filter
Price (USD)
155.40
Your Price:
Availability
***
Full specifications
Quantity
Certifications/ComplianceTarget Polypropylene Syringe Filter Housings Meet The Requirements of 21 CFR 177.1520.
DescriptionTarget2 Polypropylene Syringe Filters 0.2 µm, 4 mm, 100 Pk.
DisposableYes
MaterialPolypropylene
Material (Membrane)Polypropylene
Pore Size0.2 μm
SterilityNon-sterile
For Use With (Application)HPLC, SPE
Hold up Volume (After Air Purge)<15 μL
Housing MaterialMedical Grade, Solvent-Resistant, Virgin Polypropylene
PrefilterNo
Pressure Rating75 psi
Product LineTarget2
TypeSyringe Filter
Diameter (Metric)4 mm
Diameter (Metric) Membrane4 mm
Unit SizePack of 100
Catalog Number
Unit Size
Pack of 100
Diameter (Metric)
4 mm
Pore Size
0.45 μm
Type
Syringe Filter
Price (USD)
158.55
Your Price:
Availability
***
Full specifications
Quantity
Certifications/ComplianceTarget Polypropylene Syringe Filter Housings Meet The Requirements of 21 CFR 177.1520.
DescriptionTarget2 Polypropylene Syringe Filters 0.45 µm, 4 mm, 100 Pk.
DisposableYes
MaterialPolypropylene
Material (Membrane)Polypropylene
Pore Size0.45 μm
SterilityNon-sterile
For Use With (Application)HPLC, SPE
Hold up Volume (After Air Purge)<15 μL
Housing MaterialMedical Grade, Solvent-Resistant, Virgin Polypropylene
PrefilterNo
Pressure Rating75 psi
Product LineTarget2
TypeSyringe Filter
Diameter (Metric)4 mm
Diameter (Metric) Membrane4 mm
Unit SizePack of 100
Catalog Number
Unit Size
Pack of 100
Diameter (Metric)
17 mm
Pore Size
0.2 μm
Type
Syringe Filter
Price (USD)
333.90
Your Price:
Availability
***
Full specifications
Quantity
Certifications/ComplianceTarget Polypropylene Syringe Filter Housings Meet The Requirements of 21 CFR 177.1520.
ColorRoyal Blue
DescriptionTarget2 Polypropylene Syringe Filters 0.2 µm, 17 mm, 100 Pk.
DisposableYes
MaterialPolypropylene
Material (Membrane)Polypropylene
Pore Size0.2 μm
SterilityNon-sterile
For Use With (Application)HPLC, SPE
Hold up Volume (After Air Purge)<35 μL
Housing MaterialMedical Grade, Solvent-Resistant, Virgin Polypropylene
PrefilterNo
Pressure Rating115 psi
Product LineTarget2
TypeSyringe Filter
Volume (Metric)<29 μL (Hold Up)
Diameter (Metric)17 mm
Diameter (Metric) Membrane17 mm
Unit SizePack of 100
Showing 7 of 7
Catalog NumberSpecificationsUnit SizeDiameter (Metric)Pore SizeTypePrice (USD)AvailabilityQuantity
F2513-9Full specifications
Pack of 10017 mm0.45 μmSyringe Filter
343.35
Your Price:
***
Certifications/ComplianceTarget Polypropylene Syringe Filter Housings Meet The Requirements of 21 CFR 177.1520.
ColorWhite
DescriptionTarget2 Polypropylene Syringe Filters 0.45 µm, 17 mm, 100 Pk.
DisposableYes
MaterialPolypropylene
Material (Membrane)Polypropylene
Pore Size0.45 μm
SterilityNon-sterile
For Use With (Application)HPLC, SPE
Hold up Volume (After Air Purge)<35 μL
Housing MaterialMedical Grade, Solvent-Resistant, Virgin Polypropylene
PrefilterNo
Pressure Rating115 psi
Product LineTarget2
TypeSyringe Filter
Volume (Metric)<29 μL (Hold Up)
Diameter (Metric)17 mm
Diameter (Metric) Membrane17 mm
Unit SizePack of 100
F2500-10Full specifications
Pack of 10030 mm0.2 μmSyringe Filter
397.95
Your Price:
***
Certifications/ComplianceTarget Polypropylene Syringe Filter Housings Meet The Requirements of 21 CFR 177.1520.
ColorRoyal Blue
DescriptionTarget2 Polypropylene Syringe Filters 0.2 µm, 30 mm, 100 Pk.
DisposableYes
MaterialPolypropylene
Material (Membrane)Polypropylene
Pore Size0.2 μm
SterilityNon-sterile
For Use With (Application)HPLC, SPE
Hold up Volume (After Air Purge)<157 μL
Housing MaterialMedical Grade, Solvent-Resistant, Virgin Polypropylene
PrefilterNo
Pressure Rating150 psi
Product LineTarget2
TypeSyringe Filter
Volume (Metric)<137 μL (Hold Up)
Diameter (Metric)30 mm
Diameter (Metric) Membrane30 mm
Unit SizePack of 100
F2500-9Full specifications
Pack of 10030 mm0.45 μmSyringe Filter
447.30
Your Price:
***
Certifications/ComplianceTarget Polypropylene Syringe Filter Housings Meet The Requirements of 21 CFR 177.1520.
ColorWhite
DescriptionTarget2 Polypropylene Syringe Filters 0.45 µm, 30 mm, 100 Pk.
DisposableYes
MaterialPolypropylene
Material (Membrane)Polypropylene
Pore Size0.45 μm
SterilityNon-sterile
For Use With (Application)HPLC, SPE
Hold up Volume (After Air Purge)<157 μL
Housing MaterialMedical Grade, Solvent-Resistant, Virgin Polypropylene
PrefilterNo
Pressure Rating150 psi
Product LineTarget2
TypeSyringe Filter
Volume (Metric)<137 μL (Hold Up)
Diameter (Metric)30 mm
Diameter (Metric) Membrane30 mm
Unit SizePack of 100
F2502-9Full specifications
Pack of 10030 mm0.45 μmSyringe Filter with Pre-filter
383.25
Your Price:
***
Certifications/ComplianceTarget Polypropylene Syringe Filter Housings Meet The Requirements of 21 CFR 177.1520.
ColorWhite
DescriptionTarget2 Polypropylene Syringe Filters 0.2 µm, 30 mm, with Pre-Filter, 100 Pk.
DisposableYes
MaterialPolypropylene
Material (Membrane)Polypropylene
Pore Size0.45 μm
SterilityNon-sterile
For Use With (Application)HPLC, SPE
Hold up Volume (After Air Purge)<157 μL
Housing MaterialMedical Grade, Solvent-Resistant, Virgin Polypropylene
PrefilterYes
Pressure Rating150 psi
Product LineTarget2
TypeSyringe Filter with Pre-filter
Volume (Metric)<137 μL (Hold Up)
Diameter (Metric)30 mm
Diameter (Metric) Membrane30 mm
Unit SizePack of 100
F2504-10Full specifications
Pack of 1004 mm0.2 μmSyringe Filter
155.40
Your Price:
***
Certifications/ComplianceTarget Polypropylene Syringe Filter Housings Meet The Requirements of 21 CFR 177.1520.
DescriptionTarget2 Polypropylene Syringe Filters 0.2 µm, 4 mm, 100 Pk.
DisposableYes
MaterialPolypropylene
Material (Membrane)Polypropylene
Pore Size0.2 μm
SterilityNon-sterile
For Use With (Application)HPLC, SPE
Hold up Volume (After Air Purge)<15 μL
Housing MaterialMedical Grade, Solvent-Resistant, Virgin Polypropylene
PrefilterNo
Pressure Rating75 psi
Product LineTarget2
TypeSyringe Filter
Diameter (Metric)4 mm
Diameter (Metric) Membrane4 mm
Unit SizePack of 100
F2504-9Full specifications
Pack of 1004 mm0.45 μmSyringe Filter
158.55
Your Price:
***
Certifications/ComplianceTarget Polypropylene Syringe Filter Housings Meet The Requirements of 21 CFR 177.1520.
DescriptionTarget2 Polypropylene Syringe Filters 0.45 µm, 4 mm, 100 Pk.
DisposableYes
MaterialPolypropylene
Material (Membrane)Polypropylene
Pore Size0.45 μm
SterilityNon-sterile
For Use With (Application)HPLC, SPE
Hold up Volume (After Air Purge)<15 μL
Housing MaterialMedical Grade, Solvent-Resistant, Virgin Polypropylene
PrefilterNo
Pressure Rating75 psi
Product LineTarget2
TypeSyringe Filter
Diameter (Metric)4 mm
Diameter (Metric) Membrane4 mm
Unit SizePack of 100
F2513-10Full specifications
Pack of 10017 mm0.2 μmSyringe Filter
333.90
Your Price:
***
Certifications/ComplianceTarget Polypropylene Syringe Filter Housings Meet The Requirements of 21 CFR 177.1520.
ColorRoyal Blue
DescriptionTarget2 Polypropylene Syringe Filters 0.2 µm, 17 mm, 100 Pk.
DisposableYes
MaterialPolypropylene
Material (Membrane)Polypropylene
Pore Size0.2 μm
SterilityNon-sterile
For Use With (Application)HPLC, SPE
Hold up Volume (After Air Purge)<35 μL
Housing MaterialMedical Grade, Solvent-Resistant, Virgin Polypropylene
PrefilterNo
Pressure Rating115 psi
Product LineTarget2
TypeSyringe Filter
Volume (Metric)<29 μL (Hold Up)
Diameter (Metric)17 mm
Diameter (Metric) Membrane17 mm
Unit SizePack of 100
Showing 7 of 7
Target2 Syringe Filters are an ideal choice for general purpose laboratory use. Robust and reliable, they deliver excellent performance at an attractive price.

Outstanding Performance
  • Low extractable membranes and housings ensure that no filter-borne contaminants are added to your sample
  • Performance-tested under actual HPLC conditions
  • 30mm products are pressure rated to 120 psi
  • Housings are manufactured from solvent-resistant, low-extractable polypropylene resins specifically selected for wide compatibility with common HPLC sample matrices
  • Able to filter solutions at temperatures up to 100°C
Convenient and Easy to Use
  • Syringe filters can be sterilized by autoclave at 125°C for 15 minutes
  • Enhanced Luer Lock inlet prevents leakage; outlet fitting is a standard size male Luer-slip fitting for ease of filtrate collection
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Figures