Product Image
Invitrogen™

FISH Tag™ RNA Kits with Alexa Fluor™ Dyes

FISH Tag RNA kits provide a complete solution for fluorescence in situ hybridization (FISH) applications. Choose from kits using Alexa Fluor 488, Alexa Fluor 594, or our multicolor dye kit for probe synthesis, purification, and labeling of specimens for imaging.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLabel or DyeColor
F32952Alexa Fluor 488Green
F32954Alexa Fluor 594Red
F32956Alexa Fluor 488, Alexa Fluor 555, Alexa Fluor 594, Alexa Fluor 647Green, Red, Orange, Deep-Red
Catalog number F32952
Price (USD)
649.65
Online exclusive
866.00
Save 216.35
Each
Label or Dye:
Alexa Fluor 488
Color:
Green
Price (USD)
649.65
Online exclusive
866.00
Save 216.35
Each

Choose from two, single-color FISH Tag RNA kits with Alexa Fluor dye options (green or red). The FISH RNA multicolor kit contains four spectrally distinct dyes (green, orange, red, and deep-red) to allow simultaneous localization of multiple sequence-specific RNA and DNA targets such as mRNAs in cells, tissues, and whole embryo mounts. All kits include required reagents and come with a comprehensive, detailed protocol to help ensure your success.

Complete workflow solution for FISH

FISH Tag kits employ a two-step labeling technology. First, in vitro transcription is used to enzymatically incorporate an amine-modified nucleotide into the probe template. The modified nucleotide is UTP with an NH2 group attached through a linker to the C5 position of the base. Next, dye labeling of the purified amine-modified RNA is achieved by incubation with amine-reactive dyes (e.g., Alexa Fluor dyes). These active ester compounds react with the primary amines incorporated into the probe template, covalently conjugating the dye to the modified nucleotide base. The purified probe is then ready for hybridization to the specimen. Once the hybridization assay is complete, the specimen is viewed under a fluorescence microscope to visualize the hybridized fluorescent probe.

The dyes available in the FISH Tag RNA kits are compatible with standard filter sets found on most fluorescence microscopes. Alexa Fluor dyes are brighter and more photostable than traditional fluorescent labels, providing higher resolution and improved signal-to-noise ratios compared to conventional dyes. PureLink nucleic acid purification technology is used for fast and efficient purification of the labeled probe while SlowFade Gold Antifade Mountant preserves the fluorescent signal during imaging.

See the user manual for information regarding how to calculate base:dye ratio and nucleic acid concentration, how to construct plasmids using the FISH Tag RNA kits, and about optional RNA probe fragmentation for RNA probes greater than 500 bases.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
ColorGreen
Excitation/Emission492/520 nm
For Use With (Application)FISH Tag, fluorescent microscopy, RNA In situ hybridization
No. of Reactions10 Reactions
Product LineAlexa Fluor, FISH Tag
Product TypeRNA In Situ Hybridization Assay Kit
Quantity1 kit
Shipping ConditionDry Ice
TargetRNA
Target SpecificityNot Target-Specific
Detection MethodFluorescence
Final Product TypeProbes (Labeled RNA), Probes (Labeled DNA), Oligos (Labeled)
FormatKit
Labeling TargetRNA (General), siRNA
Label or DyeAlexa Fluor 488
Sample TypeRNA, Tissue (FFPE), Cell Culture Cells, mRNA
Unit SizeEach
Contents & Storage
Store in freezer (-5°C to -30°C) and protect from light.

Frequently asked questions (FAQs)

For imaging, what are the limitations of FISH probes?

FISH probes are generally from 300 to 1500 bp in length. To adequately image a gene or other target, the FISH probes must cover a contiguous length of ˜10 kbp. Shorter strand lengths or low-abundance DNA or RNA targets may be detected with the use of fluorescent amplification methods, such as tyramide signal amplification (TSA). For more information using TSA to amplify FISH detection, see this manual (http://tools.thermofisher.com/content/sfs/manuals/mp20912.pdf).

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

When should I use an RNA FISH probe versus a DNA FISH probe?

RNA FISH probes are the best option for hybridization to mRNA, rRNA, or other RNA targets. DNA FISH probes can also be used to detect RNA targets, but they do not hybridize as well to mRNA and other RNA target molecules. DNA FISH probes are routinely used to detect gene loci on chromosomes. RNA FISH probes can also be used to detect genes on chromosomes, but DNA FISH probes are more robust for this application.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.