<i>Funga</i>Light&trade; Yeast CFDA, AM/Propidium Iodide Vitality Kit
Invitrogen™

FungaLight™ Yeast CFDA, AM/Propidium Iodide Vitality Kit

FungaLight™ Yeast CFDA, AM/Propidium Iodide Vitality Kit
Have Questions?
Catalog NumberQuantity
F34953200 kit
Catalog number F34953
Price (MXN)
-
Quantity:
200 kit

The FungaLight™ Yeast CFDA, AM / Propidium Iodide Vitality Kit combines a cell permeable esterase substrate with a membrane integrity indicator to evaluate the vitality of yeast cells by flow cytometry or microscopy. The acetoxymethyl ester (AM) of the esterase substrate 5-carboxyfluorescein diacetate (CFDA) allows this reagent to permeant cell membranes Once inside the cell, the lipophilic blocking and diacetate groups are cleaved by nonspecific esterases, resulting in a fluorescent, charged form that leaks out of cells very slowly. In contrast, the membrane integrity indicator, propidium iodide penetrates yeast with damaged membranes. With an appropriate mixture of the CFDA, AM and propidium iodide stains, esterase-active yeast with intact cell membranes stain fluorescent green, whereas yeast with damaged membranes stain fluorescent red.

For Research Use Only. Not for use in diagnostic procedures.

Specifications
Cell TypeYeast Cells
Detection MethodFluorescence
Dye TypeOther Label(s) or Dye(s)
Emission WavelengthPI: 490/635, CFDA: 492/517
FormSolid
FormatTube(s), Slide(s)
Quantity200 kit
Shipping ConditionRoom Temperature
SolubilityDMSO (Dimethylsulfoxide)
For Use With (Application)Viability Assay
For Use With (Equipment)Fluorescence Microscope, Flow Cytometer
Product LineFungaLight
Product TypeAM/Propidium Iodide Vitality Kit
Unit SizeEach
Contents & Storage
Contains 3 vials of 5-CFDA (1 mg per vial), 1 vial of propidium iodide, and 1 vial of DMSO. Store in freezer (-5 to -30°C) and protect from light.

Frequently asked questions (FAQs)

Can I look at bacteria and yeast on a flow cytometer?

Yes, you can. We offer:

-Counting assays: Bacteria Counting Kit, for flow cytometry (Cat. No. B7277) or LIVE/DEAD BacLight Bacterial Viability and Counting Kit, for flow cytometry (Cat. No. L34856).
-Viability/vitality assays: LIVE/DEAD BacLight Bacterial Viability Kit (Cat. No. L13152).
-Membrane potential: BacLight Bacterial Membrane Potential Kit (Cat. No. B34950).
-Yeast viability/vitality assays: LIVE/DEAD FungaLight Yeast Viability Kit, for flow cytometry (Cat. No. L34952), FungaLight Yeast CFDA, AM/Propidium Iodide Vitality Kit (Cat. No. F34953)

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.