Do you have any slides of pre-labeled cells or tissues that I can use to demonstrate with?
Some of the most popular are the FluoCells demonstration slides, which have cells or tissue sections that are labeled with three different dyes. FluoCells 1 and 2 have cultured cells, while FluoCells 3 is a tissue section (mouse kidney). Slides with fluorescent beads are also available.