Emission filters for Thermo Scientific Varioskan™ LUX Multimode Microplate Readers
Actual product may vary
Emission filters for Thermo Scientific Varioskan™ LUX Multimode Microplate Readers
Emission filters for Thermo Scientific Varioskan™ LUX Multimode Microplate Readers
Thermo Scientific™

Emission filters for Thermo Scientific Varioskan™ LUX Multimode Microplate Readers

Varioskan LUX multimode microplate reader can be equipped with a range of measurement technologies including absorbance, fluorescence intensity, luminescence, AlphaScreen and time-resolved fluorescence. Additional emission filters expand the instrument functionality and application flexibility.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberWavelengthFor Use With (Application)
F51060510 nmChroma-glo luciferase CBG68 or CBG99 assay
F46080460 nmNanoBRET donor
F48020480 nmBRET donor using wtRluc
F49210492 nmGeneral Terbium measurement
F51530515 nmBRET2 acceptor using GFP2
F52010520 nmLanthaScreen measurement of Tb acceptor; HTRF donor measurement of TbGreen
F52530525 nmPierce green Renilla luciferase assay
F53030530 nmBRET acceptor using eYFP
F57177571 nmAlphaScreen measurement
F61060610 nmChroma-glo luciferase CBR assay
F610LP610 nmNanoBRET acceptor
F61520615 nmAlphaLisa measurement
F62010620 nmGeneral Europium measurement; LanthaScreen measurement of Eu donor; HTRF acceptor measurement of TbGreen; HTRF donor measurement of EuRed and TbRed
F640LP640 nmPierce red firefly luciferase assay
F66510665 nmLanthaScreen measurement of Eu acceptor; HTRF acceptor measurement of EuRed and TbRed
F47040470 nmPierce Cypridina and Gaussia luciferase assays
Catalog number F51060
Price (ZAR)
-
Wavelength:
510 nm
For Use With (Application):
Chroma-glo luciferase CBG68 or CBG99 assay

Varioskan LUX comes equipped with a range of measurement technologies including absorbance and fluorescence intensity with optional luminescence, AlphaScreen and TRF. The instrument selects the measurement wavelength using monochromators or optical filters, depending on which is optimal for each measurement technology. A wide range of optional filters extends the application area remarkably, including luciferase assays, Bioluminescence Resonance Energy Transfer (BRET) assays, Homogeneous Time Resolved Fluorescence (HTRF) assays and many other.

  • Extensive range of different type of emission filters are available for various applications when performing luminometric, TRF or Alpha technology assays, such as TR-FRET, hTRF, BRET, NanoBRET, AlphaScreen/AlphaLisa and all multiplexing assays using these detection technologies.
  • Excitation filters for TRF and AlphaScreen/AlphaLisa measurements are permanently fixed inside the corresponding detection technology optical module.
  • Emission filter installation is effortless. The instrument filter wheel is easy to access, and the correct parameters are quickly set in simple workflow in Thermo Scientific SkanIt software. SkanIt software controlled installation process also prevents accidental mistakes during the process and therefore ensures proper operation.
  • In most luminescence measurements, there is only one source of light in the system and therefore the most efficient detection is received by collecting all the light emitted by the sample, independent on the wavelength. Therefore, these assays do not require any wavelength selection. However, certain in applications, like BRET or any luminometric multiplexing assay, where multiple light sources are present in the sample, filters are used to collect the light produced at specific wavelength.
  • All Varioskan LUX filters can be used for any filter based detection technology (luminescence, TRF or Alpha technology), depending on the application. Please contact technical support for more information concerning your specific application requirements.
Specifications
Excitation/Emission510/60 nm
For Use With (Application)Chroma-glo luciferase CBG68 or CBG99 assay
Product LineVarioskan
Wavelength510 nm
For Use With (Equipment)Varioskan LUX Microplate Reader
Product TypeEmission Filter
Unit SizeEach