Notice: This product will be discontinued on May1, 2017. Please contact Customer Service for possible alternatives.

DyNAzyme Buffer (10X), Mg-free

Catalog number: F510L

Thermo Scientific™  Related applications: PCR

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Optimized Thermo Scientific 10X DyNAzyme buffers are available in several formats, with and without detergents, as well as in Mg2+-free solution.

Mg-free 10X DyNAzyme Buffer (F-510L):

• 1X buffer contains: 10 mM Tris-HCl (pH 8.8 at 25°C), 50 mM KCl, and 0.1% Triton X-100
50 mM MgCl2 is provided in a separate vial.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Polymerase: DyNAzyme II DNA Polymerase
Product Line: DyNAzyme
Product Size: 4 x 1.5 mL

Contents & storage

Contains:
• 4 x 1.5 mL 10X DyNAzyme Buffer, Mg-free
• 2 x 1.5 mL MgCl2 (50 mM)

Store at -20°C.

Documents

Manuals & protocols