Product Image
Thermo Scientific™

Phusion™ HF and GC Buffer Packs

5X Phusion Green buffers include a density reagent and two tracking dyes for direct loading of PCR products on a gel.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoNombre del producto químico o materialColor
F520LDetergent-free FF Buffer PackColorless
F518LHF Buffer PackColorless
F519LGC Buffer PackColorless
F521LDetergent-free GC Buffer PackColorless
F538LHF Buffer PackGreen
F539LGC Buffer PackGreen
Número de catálogo F520L
Precio (MXN)
-
Nombre del producto químico o material:
Detergent-free FF Buffer Pack
Color:
Colorless
Pedido a granel o personalizado
The error rate of Phusion DNA Polymerase in HF Buffer (4.4 x 10-7) is lower than that in GC Buffer (9.5 x 10-7). Therefore, the HF Buffer should be used as the default buffer for high-fidelity amplification. However, GC Buffer can improve the performance of Phusion DNA Polymerase on some difficult or long templates, such as GC-rich templates or those with complex secondary structures. For applications such as microarray or DHPLC, where the DNA templates need to be free of detergents, we recommend using detergent-free reaction buffers.

5X Phusion Green Buffers include a density reagent and two tracking dyes for direct loading of PCR products on a gel. The colored buffers do not interfere with the enzyme performance and are compatible with downstream applications such as DNA sequencing, ligation and restriction digestion.

Two optimized buffers are available for Thermo Scientific Phusion High-Fidelity DNA Polymerases:

  • 5X Phusion HF Buffer (F-518L, F-538L (Phusion Green), F-520L (Detergent-free))
  • 5X Phusion GC Buffer (F-519L, F-539L (Phusion Green), F-521L (Detergent-free))
Para uso exclusivo en investigación. No apto para uso en procedimientos diagnósticos.
Especificaciones
Nombre del producto químico o materialDetergent-free FF Buffer Pack
ColorColorless
DescripciónPaquete de tampones HF Phusion™ libre de detergente
IncluyeEl paquete de tampones HF Phusion, sin detergentes (F-518L) incluye:
• Tampón HF 5X Phusion sin detergentes (el tampón 1X proporciona 1,5 mM de MgCl2)
• 50 mM de MgCl2
• DMSO

Almacenar a -20 °C.
Para utilizar con (aplicación)Standard PCR
Cantidad4 x 1,5 mL
Unit SizeEach