Thermo Scientific™
Thermolyne™ Atmosphere Controlled Ashing Furnaces
 Related applications:
Lab Ovens Furnaces
Achieve consistent temperature control with the Thermo Scientific™ Thermolyne™ Atmosphere Controlled Ashing Furnace, which reaches 975°C with the standard stainless-steel manifold and 1093°C with the optional inconel manifold.
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Back to top
Description
Key Features:
  • Adjustable gas flowmeter/valve (0-80L/min.) on front for easy access when adjusting the airflow rate
  • Stainless-steel manifold at rear chamber prewarms incoming gases, provides a maximum temperature gradient of only ±3°C at 750°C
  • Chamber rear has a 0.95cm (0.38in.) port for monitoring chamber temperatures with independent measuring devices
  • With 0.64cm (0.25in.) I.D. or 0.96cm (0.375in.) O.D. hose barb (in chamber rear) for inert gas line
Type F6000:
  • With two dual-purpose stainless-steel trays and handle to accommodate 24 (30mL) porcelain crucibles or 38 (10mL) quartz crucibles
Type F6000-60:
  • Meets ASTM™ D3174 specifications: 3 to 4 air exchanges per min.
  • Heating rate of 8°C/min. to 500°C, 6°C/min. from 500° to 750°C
  • Holds at 750°C for two hours, then turns off
Requires:
Electrical connection requires fixed wiring (power cord not provided).
Compatible with:
  • Stainless-steel shelf (SH408X1)
  • Stainless-steel flexible exhaust tubing kit with mounting hardware (AY408X1A)
Documents
Manuals & protocols