Search
View additional product information for Phusion™ Plus DNA Polymerase - FAQs (F630S, F630XL, F630L)
2 product FAQs found
Please see "Conclusions" within the technical note for the Phusion Plus DNA Polymerase (Cat. No. F630S) (see link: https://assets.thermofisher.com/TFS-Assets/BID/Technical-Notes/measuring-error-rates-ngs-dna-polymerases-tech-note.pdf).
Find additional tips, troubleshooting help, and resources within our PCR and cDNA Synthesis Support Center.
Reaction setup with Phusion Plus DNA Polymerase or Phusion Plus PCR Master Mix can be done either on ice or at room temperature.