Cyto-Cal™ Multifluor + Violet Intensity Calibrator
Thermo Scientific™

Cyto-Cal™ Multifluor + Violet Intensity Calibrator

Simultaneously monitor stability and check sensitivity and performance of flow cytometer.
Have Questions?
Número de catálogoCantidad
FC3MV1,5 x 10 partículas/mL
Número de catálogo FC3MV
Precio (MXN)
-
Cantidad:
1,5 x 10 partículas/mL
Simultaneously monitor stability and check sensitivity and performance of flow cytometer with the Thermo Scientific™ Cyto-Cal™ Multifluor + Violet Intensity Calibrator.
Features:
  • Stable calibration material for assessing instrument stability and precision in multiple channels
  • Uniform mixture of 3μm particles with four dyes in five different fluorescent intensities
  • Singlet gating not required due to high uniformity of the particles
  • Consists of a single vial of fluorescent particles precisely stained with fluorescent dyes that have optimized intensity and broad emission in up to 13 color channels
  • MESF (Mean Equivalent Soluble Fluorochrome) value provided for each intensity level for estimation of relative fluorescence intensities of labeled cells
  • Can provide relative fluorescence estimations of labeled cells (MEFL), but is not designed for quantitation of fluorochromes on cells.

Includes:

Insert sheet and MSDS

Recommended for:

Flow Cytometry set-up and calibration.

Note:

MEFL is also referred to as Molecules of Equivalent Soluble Fluorochrome (MESF). The MEFL values provided are not traceable to a standard reference material.

Especificaciones
Aditivo0.05% Tween-20 Dispersant / Surfactant with 2 mM Sodium Azide preservative
DescripciónBeads for assessing instrument stability and precision in multiple channels
Diámetro (métrico)3 μm
Tipo de coloranteFirefli™ Fluorescent Green (488/510nm), Orange (488/575nm), Red (488, 633, 635/700nm) and Blue (405/
IncluyeDyed polymer microspheres in water, Package insert sheet
Nivel1 blank and 5 fluorescent
MaterialPoliestireno
Nº de pruebas50
Densidad de las partículas1.06g/cm3
Cantidad1,5 x 10 partículas/mL
EstabilidadStable for a minimum of 12 months
Uniformidad<3%
Volumen (métrico)2 mL
Línea de productosCyto-Cal
Tipo de productoInstrument Calibration
Unit SizeEach
Contenido y almacenamiento
Unless otherwise stated, refrigerate (2° to 8°C) product when not in use but do not freeze. Store upright and keep bottle tightly sealed. Mix product with gentle inversion by hand or vortex mixer.