FDP120 Oil Vacuum Pumps
Actual product may vary
Thermo Scientific™
FDP120 Oil Vacuum Pumps
Use specifically designed Thermo Scientific™ FDP120 Oil Vacuum Pumps for standard freeze drying applications.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVoltage
FDP120-230230 V
FDP120-115115 V
Catalog number FDP120-230
Price (MXN)
-
Voltage:
230 V
Use specifically designed Thermo Scientific™ FDP120 Oil Vacuum Pumps for standard freeze drying applications.
  • 116L/min (at 60Hz) throughput
  • 1.5mtorr ultimate vacuum level
  • Supplied Gas Ballast Oil Return Kit (GBORK) allows the user to apply full gas ballast to keep the oil clean without losing oil
  • Oil mist filter filters the exhaust to keep environmental air clean
Ordering Information:
For applications involving aggressive solvents that might damage ordinary pumps, we recommend the LyoPump VLP195FD which also includes an active oil filtration system (VPOF110) that dramatically extends the life of the pump.
Includes:
GBORK (Gas Ballast Oil Return Kit) and Oil Mist Filter (EMF-10).
Specifications
ApplicationsStandard Freeze Drying
Electrical Requirements230 V 50 Hz
Frequency50 Hz
Product LineThermo Scientific
Flow Rate116 L/min
Max. Vacuum1.5 mtorr
Pump TypeVacuum Pump
Voltage230 V
Unit SizeEach