Specifications
Amperage13.5 A
Breaker20A
Cabinet MaterialScratch-resistant Painted
Control TypeMicroprocessor
DefrostManual
Depth (English) Interior29 in.
Depth (Metric) Interior74 cm
Dimensions (D x W x H) Interior29 x 30 x 58 in. (74 x 76 x 147 cm)
Dimensions (L x W x H) Exterior37.2 x 34 x 79.2 in. (94.5 x 86.4 x 201 cm)
Door StyleSolid
Electrical Requirements115 V 60 Hz
Exterior FinishEpoxy Coated Steel
Frequency60 Hz
Height (English) Exterior79.2 in.
Height (English) Interior58 in.
Height (Metric) Exterior201 cm
Height (Metric) Interior147 cm
InsulationUrethane Foam
Length (English) Exterior37.2 in.
Length (Metric) Exterior94.5 cm
Plug TypeNEMA 5-15 P
Rack Capacity9
RefrigerantCFC-free
Shipping Weight (English)445 lb.
Shipping Weight (Metric)197 kg
TypeEnzyme Freezer
WarrantyWarranty varies by country, please contact us for details
Width (English) Exterior34 in.
Width (English) Interior30 in.
Width (Metric) Exterior86.4 cm
Width (Metric) Interior76 cm
Capacity (English)29.2 cu. ft.
Capacity (Metric)826 L
Certifications/ComplianceUL
CompressorHeavy-duty, Industrial-grade, Hermetic
Monitoring OptionsAlarm
No. of Doors1
Shelves9
Temperature Range-20°C
Voltage115 V
Unit SizeEach