Forma™ High-Performance Auto Defrost Lab Freezers
Forma™ High-Performance Auto Defrost Lab Freezers
Actual product may vary
Thermo Scientific™

Forma™ High-Performance Auto Defrost Lab Freezers

Protect sensitive medical and scientific storage with the advanced features of this high-performance lab freezer, designed for storage of common laboratory materials.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Electrical RequirementsPlug Type
FFGL1230V326 L230 V 50 HzEU
FFGL430A133 L115 V 60 HzNEMA 5-15 P
FFGL1230A326 L115 V 60 HzNEMA 5-15 P
FFGL2330A659 L115 V 60 HzNEMA 5-20
FFGL2330D659 L208 - 230 V 60 Hz 6-15 PNEMA 6-15 P
FFGL2330V659 L230 V 50 HzEU
FFGL3030A826 L115 V 60 HzNEMA 5-20 P
FFGL3030D826 L208 - 230 V 60 HzNEMA 6-15 P
FFGL3030V826 L230 V 50 HzEU
FFGL5030A1447 L115 V 60 HzNEMA 5-20 P
FFGL5030D1447 L208 - 230 V 60 Hz 6-15 PNEMA 6-15 P
FFGL5030V1447 L230 V 50 HzEU
Catalog number FFGL1230V
Price (BRL)
-
Capacity (Metric):
326 L
Electrical Requirements:
230 V 50 Hz
Plug Type:
EU
Protect sensitive medical and scientific storage with the advanced features of Thermo Scientific™ Forma™ High-Performance Lab Freezers, designed for medical and scientific applications such as storage of reagents, pharmaceuticals, biologicals and other commonly used laboratory materials. Our Forma high-performance freezers feature microprocessor control, positive airflow systems, industrial-quality cabinet construction and extra-strength refrigeration compressors selected for industrial, clinical and scientific use.
Key Features:
  • Stable, low-temperature environment for frequently accessed laboratory reagents, pharmaceuticals and biological materials
  • Temperature factory-preset at -30°C†
  • Automatic defrost and condensate removal

Microprocessor Control:

  • Easy-to-use controls and visual displays
  • Alarm silence, ringback and automatic reset functions
  • Setpoint security with key-operated, triple-position master switch that controls power to all systems
  • Audible/visual alarms for high/low temperature and power failure
  • Battery backup and remote alarm connections

Construction:

  • Rugged, long-life cabinets with smooth, scratch-resistant interiors‡ and exteriors that are easy to clean
  • High-density, CFC-free insulation
  • Solid, self-closing doors with keyed lock
  • Closed-cell gasket provides a positive door seal to maintain a constant interior temperature
  • Directed airflow for fast temperature recovery after door openings
  • Full-length door handles
  • Painted open-wire shelves are adjustable in 1 in. (2.5cm) increments

Refrigeration System:

  • Heavy-duty, industrial-grade, hermetically sealed compressors
  • Automatic defrost maintains optimum cooling

Ordering Information:

  • Options include: freestanding low-voltage booster; built-in temperature chart recorder; 2in. (5cm) media/probe port, with cover; stainless-steel interiors and exteriors; 2in. (5cm) casters; and extra shelves. V and W models are not
  • Available in North America. Contact your customer service or sales representative for more information.

Note:

†Performance may be affected by ambient temperature and humidity conditions, sample loading preferences, and frequency of door openings.

‡Specified interior volumes are nominal, and usable capacities may be offset by shelves, drawers, blower coils or cabinet protrusions.

Warranty varies by country, please contact us for details
Specifications
Amperage8 A
Breaker15A
Control TypeMicroprocessor
DefrostAutomatic
Depth (English) Interior21.75 in.
Depth (Metric) Interior50.8 cm
Dimensions (D x W x H) Interior21.75 x 20 x 52.4 in. (50.8 x 50.8 x 133.1 cm)
Dimensions (L x W x H) Exterior29.3 x 24 x 73.6 in. (74.3 x 61 x 186.9 cm)
Electrical Requirements230 V 50 Hz
Exterior FinishStainless Steel
Frequency50 Hz
Height (English) Exterior73.6 in.
Height (English) Interior52.4 in.
Height (Metric) Exterior186.9 cm
Height (Metric) Interior133.1 cm
InsulationCFC-free
Length (English) Exterior29.3 in.
Length (Metric) Exterior74.3 cm
RefrigerantCFC-free
Shipping Weight (English)280 lb.
TypeHigh-Performance Freezer
WarrantyWarranty varies by country, please contact us for details
Width (English) Exterior24 in.
Width (English) Interior20 in.
Width (Metric) Exterior61 cm
Width (Metric) Interior50.8 cm
Cabinet MaterialStainless Steel
Capacity (English)1.5 cu. ft.
Capacity (Metric)326 L
Certifications/ComplianceUL
CompressorHeavy-duty, Industrial-grade, Hermetic
Door StyleSolid
Monitoring OptionsAlarm, Recorder
No. of Doors1
Plug TypeEU
Shelves4
StyleSpace Saver
Temperature Range-30°C
Voltage230 V
Unit SizeEach