Forma™ High-Performance Manual Defrost Lab Freezers
Actual product may vary
Thermo Scientific™

Forma™ High-Performance Manual Defrost Lab Freezers

Protect sensitive medical and scientific storage with the advanced features of this high-performance lab freezer, designed for storage of common laboratory materials.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (English)Electrical RequirementsPlug Type
FFGL3020W29.2 cu. ft.230 V 50 HzUK
FFGL2320A23.3 cu. ft.115 V 60 HzNEMA 5-20 P
FFGL2320D23.3 cu. ft.208 - 230 V 60 HzNEMA 6-15 P
FFGL2320V23.3 cu. ft.230 V 50 HzEU
FFGL2320W23.3 cu. ft.230 V 50 HzUK
FFGL3020A29.2 cu. ft.115 V 60 HzNEMA 5-20 P
FFGL3020D29.2 cu. ft.208 - 230 V 60 HzNEMA 6-15 P
FFGL3020V29.2 cu. ft.230 V 50 HzEU
Catalog number FFGL3020W
Price (CLP)
-
Capacity (English):
29.2 cu. ft.
Electrical Requirements:
230 V 50 Hz
Plug Type:
UK
Protect sensitive medical and scientific storage with the advanced features of Thermo Scientific™ Forma™ High-Performance Lab Freezers, designed for medical and scientific applications such as storage of reagents, pharmaceuticals, biologicals and other commonly used laboratory materials. Our Forma high-performance freezers feature microprocessor control, positive airflow systems, industrial-quality cabinet construction and extra-strength refrigeration compressors selected for industrial, clinical and scientific use.
Key Features:
  • Stable, low-temperature environment for frequently accessed laboratory reagents, pharmaceuticals and biological materials
  • Temperature factory-preset at -20°C†
  • Manual defrost

Microprocessor Control:

  • Easy-to-use controls and visual displays
  • Alarm silence, ringback and automatic reset functions
  • Setpoint security with key-operated, triple-position master switch that controls power to all systems
  • Audible/visual alarms for high/low temperature and power failure
  • Battery backup and remote alarm connections

Construction:

  • Rugged, long-life cabinets with smooth, scratch-resistant interiors‡ and exteriors that are easy to clean
  • High-density, CFC-free insulation
  • Solid, self-closing doors with keyed lock
  • Closed-cell gasket provides a positive door seal to maintain a constant interior temperature
  • Directed airflow for fast temperature recovery after door openings
  • Full-length door handles
  • Painted open-wire shelves are adjustable in 1 in. (2.5cm) increments

Refrigeration System:

  • Heavy-duty, industrial-grade, hermetically sealed compressors
  • Manual defrost for maximum uniformity performance

Ordering Information:

  • Options include: freestanding low-voltage booster; built-in temperature chart recorder; 2in. (5cm) media/probe port, with cover; stainless-steel interiors and exteriors; 2in. (5cm) casters; and extra shelves. V and W models are not
  • Available in North America. Contact your customer service or sales representative for more information.

Note:

†Performance may be affected by ambient temperature and humidity conditions, sample loading preferences, and frequency of door openings.

‡Specified interior volumes are nominal, and usable capacities may be offset by shelves, drawers, blower coils or cabinet protrusions.

Warranty varies by country, please contact us for details
Specifications
Amperage12 A
Breaker15A
Control TypeMicroprocessor
DefrostManual
Depth (English) Interior30 in.
Depth (Metric) Interior76 cm
Dimensions (D x W x H) Interior30 x 29 x 58 in. (76 x 74 x 147 cm)
Dimensions (L x W x H) Exterior34 x 37.2 x 79.2 in. (86.4 x 94.5 x 201 cm)
Electrical Requirements230 V 50 Hz
Exterior FinishStainless Steel
Frequency50 Hz
Height (English) Exterior79.2 in.
Height (English) Interior58 in.
Height (Metric) Exterior201 cm
Height (Metric) Interior147 cm
InsulationCFC-free
Length (English) Exterior34 in.
Length (Metric) Exterior86.4 cm
RefrigerantCFC-free
Shipping Weight (English)446 lb.
Shipping Weight (Metric)197 kg
TypeHigh-Performance Freezer
WarrantyWarranty varies by country, please contact us for details
Width (English) Exterior37.2 in.
Width (English) Interior29 in.
Width (Metric) Exterior94.5 cm
Width (Metric) Interior74 cm
Cabinet MaterialStainless Steel
Capacity (English)29.2 cu. ft.
Capacity (Metric)826 L
CompressorHeavy-duty, Industrial-grade, Hermetic
Door StyleSolid
Monitoring OptionsAlarm, Recorder
No. of Doors1
Plug TypeUK
Shelves4
StyleUpright
Temperature Range-20°C
Voltage230 V
Unit SizeEach