Cyto-Plex™ Carboxylated Particles
Thermo Scientific™

Cyto-Plex™ Carboxylated Particles

Save time when measuring analytes with Thermo Scientific™ Cyto-Plex™ Carboxylated Particles, open platform particles for suspension array analysis.
Have Questions?
變更視圖buttonViewtableView
產品號碼Diameter (Metric)QuantityLevel
FM4CR024 μm1 mLLevel 2
FM4CR014 μm1 mLLevel 1
FM4CR01B4 μm5 mLLevel 1
FM4CR02B4 μm5 mLLevel 2
FM4CR034 μm1 mLLevel 3
FM4CR03B4 μm5 mLLevel 3
FM4CR044 μm1 mLLevel 4
FM4CR04B4 μm5 mLLevel 4
FM4CR054 μm1 mLLevel 5
FM4CR05B4 μm5 mLLevel 5
FM4CR064 μm1 mLLevel 6
FM4CR06B4 μm5 mLLevel 6
FM4CR074 μm1 mLLevel 7
FM4CR07B4 μm5 mLLevel 7
FM4CR084 μm1 mLLevel 8
FM4CR08B4 μm5 mLLevel 8
FM4CR094 μm1 mLLevel 9
FM4CR09B4 μm5 mLLevel 9
FM4CR104 μm1 mLLevel 10
FM4CR10B4 μm5 mLLevel 10
FM4CR114 μm1 mLLevel 11
FM4CR11B4 μm5 mLLevel 11
FM4CR124 μm1 mLLevel 12
FM4CR12B4 μm5 mLLevel 12
FM5CR015 μm1 mLLevel 1
FM5CR01B5 μm5 mLLevel 1
FM5CR025 μm1 mLLevel 2
FM5CR02B5 μm5 mLLevel 2
FM5CR035 μm1 mLLevel 3
FM5CR03B5 μm5 mLLevel 3
FM5CR045 μm1 mLLevel 4
FM5CR04B5 μm5 mLLevel 4
FM5CR055 μm1 mLLevel 5
FM5CR05B5 μm5 mLLevel 5
FM5CR065 μm1 mLLevel 6
FM5CR06B5 μm5 mLLevel 6
FM5CR075 μm1 mLLevel 7
FM5CR07B5 μm5 mLLevel 7
FM5CR085 μm1 mLLevel 8
FM5CR08B5 μm5 mLLevel 8
FM5CR095 μm1 mLLevel 9
FM5CR09B5 μm5 mLLevel 9
FM5CR105 μm1 mLLevel 10
FM5CR10B5 μm5 mLLevel 10
FM5CR115 μm1 mLLevel 11
FM5CR11B5 μm5 mLLevel 11
FM5CR125 μm1 mLLevel 12
FM5CR12B5 μm5 mLLevel 12
產品號碼 FM4CR02
價格 (TWD)
-
Diameter (Metric):
4 μm
Quantity:
1 mL
Level:
Level 2
Save time when measuring analytes with Thermo Scientific™ Cyto-Plex™ Carboxylated Particles, open platform particles for suspension array analysis.
Suspension array analysis is a powerful technique enabling the simultaneous detection and quantitation of multiple analytes with a single sample. Using color coded microspheres with the appropriate dyes, these tests can now be carried out using commonly available flow cytometers.
  • Cyto-Plex™ microspheres consist of a highly uniform polystyrene particle with fluorescent intensities which are completely separated from each other
  • Carboxylated particles provide different sizes and levels of fluorescent intensities for analysis of multiple analytes
  • Use of a single diameter particle for all dye levels saves time by only requiring the development and optimization of one bead chemistry; multiple diameters can be combined to increase the number of analytes measured in one test
  • High-density binding sites and low non-specific binding enable coupling with a wide variety of antibodies, nucleic acids and other biomolecules
  • Microspheres have a maximum emission at 700 nm and can be excited with either 488 nm or 633 nm lasers
  • Emission can be collected in either the PE-Cy5 or APC channels
  • Since there is little or no emission in the FITC and PE channels, probes utilizing either of these dyes can be effectively used as reporters
  • Open platform microspheres for suspension array analysis in flow cytometry
  • 規格
    AdditiveSodium Azide Preservative, 0.05% Tween-20
    ContentsPolystyrene microspheres in water
    DescriptionCyto-Plex Carboxylated Particles, 4µm
    Dye TypeFirefli™ Fluorescent Red (635/700nm)
    IncludesMaterial Safety Data Sheet (MSDS) and Package Insert Sheet
    LevelLevel 2
    Particle Density1.06g/cm³
    Quantity1 mL
    StabilityProduct stable for a minimum of 12 months
    Surface ModificationCarboxylate-Modified
    Uniformity≤2% CV
    ColorRed
    Diameter (Metric)4 μm
    MaterialPolystyrene with Copolymer Grafted Surface
    Product TypeCarboxylated Particle
    Unit SizeEach
    內容物與存放
    Refrigerate when not in use; do not freeze. Store upright and keep bottle tightly sealed. Mix product by gentle inversion by hand or vortex mixer.