Product Image

Tissue Extraction Reagent II

Lysis buffer for total protein extraction from tissue samples
Have Questions?
Catalog NumberQuantity
FNN0081100 ml
Catalog number FNN0081
Price (USD)
198.00
Each
Quantity:
100 ml
Price (USD)
198.00
Each

Build Your Own Immunoassay, Cell Analysis, Cell Lysis, Immunoassays, Protein Sample Preparation and Protein Purification, Proteins, Expression, Isolation and Analysis

Tissue Extraction Reagent II is for ready-to-use bicine-based lysis buffer for total protein extraction from tissue samples by homogenization. Cell lysates prepared with Tissue Extraction Reagent II are compatible with immunoassays such as ELISA, western blot, and Luminex assay.

Buffer formulation: 25 mM Bicine, pH 7.5, 150 mM NaCl, 2 mM Na3VO4, 1 mM NaF, 20 mM 4P2O7, 0.02% NaN3, detergent

This cell lysis buffer does not contain protease inhibitors and should be supplemented with 1 mM PMSF and protease inhibitor cocktail (Cat. No. A32955) just prior to use.

Tissue Extraction Reagent II may be supplemented with salts, reducing agents or chelating agents as needed.

Specifications
For Use With (Application)Luminex Assay, Western Blot, Sample preparation for ELISA
Product TypeTissue Extraction Reagent
Quantity100 ml
Shipping ConditionDry Ice
Unit SizeEach
Contents & Storage
Store at 2°C to 8°C

Frequently asked questions (FAQs)

What extraction reagents are recommended for efficient mouse tissue analysis?

We have 5 different cell and tissue extraction buffers suitable for preparing mouse cell and tissue extracts. These buffers can be used to extract cells and tissues from many other species as well. The exact compositions of all of our buffers are proprietary, but they are similar to those described by many researchers.

Four of these buffers can be used to prepare extracts which can be analyzed with our ELISA and Luminex kits and by Western blotting. Our Cell Extraction Buffer (FNN0011) contains extra phosphatase inhibitors and resembles the RIPA formulation that many people use. Our Tissue Extraction Reagents I (FNN0071) and II (FNN0081) contain different concentrations of NaCl and different surfactants, but are otherwise similar to each other. For those who prefer using an extraction buffer containing the detergent NP-40, we have our NP-40 Lysis Buffer (FNN0021). Finally, we sell a Denaturing Cell Extraction buffer (FNN0091) which contains 3 detergents and a chaotropic agent. Extracts prepared with FNN0091 can be analyzed with our ELISA kits and by Western blotting only. These buffers do not contain protease inhibitors, which the investigator should add right before use.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

What is the difference in application between Tissue Extraction Reagent I (Cat. No. FNN0071) and Tissue Extraction Reagent II (Cat. No. FNN0081)?

The main difference is that Tissue Extraction Reagent II is more stringent than Tissue Extraction Reagent I. Additionally, please note that Tissue Extraction Reagent I contains EDTA which may not be compatible with some downstream applications.

Find additional tips, troubleshooting help, and resources within our Antibodies and Immunoassays Support Center.

During my ProcartaPlex assay data analysis, I am getting a warning message that there is high bead aggregation. What should I do?

Here are possible causes and solutions for this issue:

- Check the protocol settings (make sure you select the correct DD settings).
- Check the level of sheath fluid and empty the waste.
- Before acquiring the plate, run calibration and verification beads on the Luminex instrument.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

During my ProcartaPlex assay data analysis, the beads fall below or to the lower left of their bead region on the bead map. Why is this?

Here are possible causes and solutions for this issue:

This usually indicates that the beads have been photobleached. This problem can also be caused by exposing the beads to organic solvents. Unfortunately, the assay will have to be repeated because the beads cannot be restored. The beads must be protected from light and organic solvents.
Alternatively, the instrument may be off in its measurements or you may have a calibration issue. Call the manufacturer for a service appointment.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

During my ProcartaPlex assay data analysis, beads do not appear in the region gated. What happened?

This indicates that an incorrect buffer was used for the final step. The Wash Solution provided in the kit must be used for washing the beads and the Reading Buffer should be used for resuspending the beads before loading them into the Luminex instrument. The osmolarity of the solution will impact the size of the bead, and any change in the bead size will alter detection by the instrument.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.