Owl™ Blotting Paper
Thermo Scientific™

Owl™ Blotting Paper

Absorb adequate amounts of buffer for semi-dry electroblotting without the loss of mechanical stability with this high-quality blotting paper.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Width (Metric)
FP-235 cm45 cm
FP-120 cm20 cm
FP-346 cm57 cm
FP-410 cm10 cm
FP-518 cm20 cm
FP-69 cm9 cm
FP-712 cm16 cm
Catalog number FP-2
Price (USD)
251.10
Online Exclusive
279.00
Save 27.90 (10%)
Pack of
Length (Metric):
35 cm
Width (Metric):
45 cm
Price (USD)
251.10
Online Exclusive
279.00
Save 27.90 (10%)
Pack of

Absorb adequate amounts of buffer for semi-dry electroblotting without the loss of mechanical stability with Thermo Scientific™ Owl™ Blotting Paper, high-quality blotting paper ideal for wicking and blotting applications.

  • High purity, 100% cotton fiber
  • Available in pre-cut mini gel and large format gel sizes
  • Compatible with Owl electroblotting systems

Ordering Information:

  • Sold in packs of 100 sheets
Specifications
Flow Rate35 mL/min.
For Use With (Equipment)Owl electroblotting systems
Packaging TypeSheets
Weight183 g/m2
Dimensions (LxW)35 cm x 45 cm
FormatSheet
Length (Metric)35 cm
Material100% Cotton Fiber
Product LineOwl
Thickness0.83 mm
Width (Metric)45 cm
Unit SizePack of