Cell Holder for Sealed Precision Pathlength Cells
Thermo Scientific™
Cell Holder for Sealed Precision Pathlength Cells
Thermo Scientific™ Cell Holder for Sealed Precision Pathlength Cells works with all Sealed Precision Pathlength Cells, regardless of pathlength or window material.
Have Questions?
Catalog NumberFor Use With (Application)Type
FT04-035Transmission liquid sampling; Quantitative analysis; Quality control analysisFT-IR Cell Holder
Catalog number FT04-035
Price (CLP)
-
For Use With (Application):
Transmission liquid sampling; Quantitative analysis; Quality control analysis
Type:
FT-IR Cell Holder
Thermo Scientific™ Cell Holder for Sealed Precision Pathlength Cells works with all Spectra-Tech Sealed Precision Pathlength Cells, regardless of pathlength or window material.
The cell holder features an even seal for leakproof operation. An amalgam seal guarantees excellent performance, even with highly volatile samples. Two PTFE stoppered ports facilitate filling and cleaning of the cell.
  • Even seal design provides even pressure, leakproof sampling, and minimized cell breakage
  • Quick-change holder for 32mm windows
  • Luer-Lok fittings for convenient filling
Includes:
Back plate, PTFE gasket, PTFE o-ring and knurled end cap
Recommended for:
  • Transmission liquid sampling
  • Quantitative analysis
  • Quality control analysis
Specifications
DescriptionThe Thermo Scientific Cell Holder for Demountable Cells.
Diameter (Metric)32 mm
For Use With (Application)Transmission liquid sampling; Quantitative analysis; Quality control analysis
IncludesBack plate, PTFE gasket, PTFE o-ring and knurled end cap.
TypeFT-IR Cell Holder
Unit SizeEach