Product Image
Thermo Scientific™
Oxoid™ Listeria Rapid Test 2.25L
Thermo Scientific™ Oxoid Listeria Rapid Test 2.25L is a rapid monoclonal antibody test for presumptive identification of Listeria spp.
Catalog NumberQuantity
FT0401A50 Tests/Kit
Catalog number FT0401A
Price (GBP)
-
Quantity:
50 Tests/Kit
Thermo Scientific™ Oxoid Listeria Rapid Test 2.25L is a rapid monoclonal antibody test for presumptive identification of Listeria spp. in food or food processing equipment.
  • Device contains specific monoclonal antibodies to the B flagella antigen common in many Listeria spp.
Includes:
50 test devices; 3 positive control; 5 vials of Half Fraser supplements; each vial sufficient for 2.25L Fraser Broth Base
Specifications
Control SetsPositive
DescriptionListeria Rapid Test 2.25L
Detectable AnalytesListeria spp. in food or food processing equipment
TypeListeria Test Kit
Quantity50 Tests/Kit
Target Organism ClassListeria Spp.
Unit SizeEach