Thermo Scientific™
Superfrost™ Plus Gold Slides
 Related applications:
Anatomical Pathology
Firmly attract and chemically bond fresh or formalin-fixed frozen tissue sections to the slide surface with Thermo Scientific™ Superfrost™ Plus Gold Slides.
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Back to top
Description
  • Specifically designed for frozen tissue sections and recommended for bone, brain and breast tissues that are 5 microns or larger
  • Ideal for special stains, immunocytochemical and in situ DNA hybridization techniques when applied to fixed or frozen tissue sections
  • Compatible with both toluidine blue and hematoxylin and eosin rapid frozen section stains
Documents
Manuals & protocols