E-Gel™ iBase™ Power System (EU only) - FAQs
E-Gel™ iBase™ Power System (EU only) - FAQs
View additional product information for E-Gel™ iBase™ Power System (EU only) - FAQs (G6400EU)
1 product FAQs found
My E-Gel iBase/PowerBase/E-Base system is continuously beeping and/or shows a steady red light. What am I doing wrong?
Check the E-Gel cassette and copper contacts. You can try replacing the gel cassette with a fresh gel cassette to see whether this fixes the problem. A cold cassette or improper operating conditions can also lead to this failure. Cassettes should be at room temperature for use; avoid storing at 4°C.
Find additional tips, troubleshooting help, and resources within our Nucleic Acid Purification and Analysis Support Center.