E-Gel™ EX Agarose Gels Basic Starter Kit, 2%, UK Version

Catalog number: G6412STUK

Invitrogen™  Related applications: Nucleic Acid Gel Electrophoresis & Blotting

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The E-Gel® EX 2% Agarose Basic Starter Kit is an economical way to get started using E-Gel® EX agarose gels. It provides:

• One box of 10 E-Gel® EX 2% Agarose Gels
E-Gel® iBase™ Power System
E-Gel® 1 Kb Plus DNA Ladder
E-Gel® Sample Loading Buffer
• One gel knife

E-Gel® EX 1% Agarose Gels are self-contained, pre-cast agarose gels designed to provide fast, convenient, and easy electrophoresis. Each E-Gel® agarose gel is ready for use with agarose, electrodes, and SYBR® Gold DNA gel stain packaged inside an 11-well, easy-to-open, UV- and blue-light transparent cassette. SYBR® Gold stain can be viewed with blue light, which minimizes DNA damage and allows real-time monitoring of DNA migration with the E-Gel® Safe Imager Real-Time Transilluminator.

Ultra-sensitive detection of DNA
E-Gel® EX gels were developed for ultimate sensitivity and demonstrate over 5-fold greater sensitivity than comparable gels containing ethidium bromide. This is due to the use of the ultra-sensitive SYBR® Gold nucleic acid gel stain. The superior sensitivity allows you to use lower amounts of sample, saving time and money.

Quick check of RNA samples
E-Gel® EX gels can also be used for fast analysis of RNA samples to check on their integrity before proceeding with downstream applications.

Easy access to the gel
The gel cassette is designed to open with a gel knife (included with this kit), so that the gel inside can be accessed readily for excision of specific bands or transferred to a membrane for Southern blot analysis.

Running and viewing apparatus
E-Gel® EX gels require the E-Gel® iBase™ Power System (included) for running and the E-Gel® Safe Imager™ Real-Time Transilluminator (not included) for real-time viewing of sample migration.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Gel Percentage: 2 %
Gel Type: E-Gel®
In-Gel Dye: SYBR® Gold II
Product Line: E-Gel®
Product Size: 1 kit
Well Format: Single Comb
Wells: wells
Shipping Condition: Room Temperature

Contents & storage

E-Gel® iBase, UK version
E-Gel® 1 Kb Plus DNA Ladder
1 box of 10 E-Gel EX gels, 2%
E-Gel® Sample Loading Buffer, 1X
1 gel knife

Store at room temperature.

Documents

Manuals & protocols

Recommended products