Invitrogen™

Safe Imager™ 2.0 Power Cord Replacement

Catalog number:  G6602

 Related applications: Nucleic Acid Gel Electrophoresis & Blotting | Protein Gel Electrophoresis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The Safe Imager™ Blue-Light Transilluminator is designed for viewing stained gels on the laboratory bench top, or within gel documentation systems. Light from the LED source inside the instrument passes through a blue filter producing light with a narrow emission peak centered at approximately 470 nm, effective for the excitation of SYBR® DNA-binding dyes such as SYBR® Safe DNA gel stain, as well as many of our protein gel stains such as SYPRO® Ruby, SYPRO® Orange, and Pro-Q® Diamond stains. Sensitivity obtained using this instrument is comparable to that obtained with a standard UV transilluminator. Unlike UV transilluminators, the Safe Imager™ transilluminator does not produce UV light and does not require UV protective equipment during use.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment): Safe Imager™
Product Line: Safe Imager™
Product Size: 1 unit
Type: Cords and Connectors
Shipping Condition: Room Temperature

Contents & storage

Each instrument comes with an amber filter unit and viewing glasses.

Documents

Manuals & protocols

Recommended products