E-Gel™ Power Snap Electrophoresis System Starter Kit, SYBR Safe 1%
Invitrogen™
E-Gel™ Power Snap Electrophoresis System Starter Kit, SYBR Safe 1%
Provides everything needed to quickly start using precast 1.2% E-Gel agarose gels with SYBR Safe on the system for a streamlined gel electrophoresis and gel imaging workflow.
Have Questions?
Número de catálogoCantidad
G8321ST1 Kit
Número de catálogo G8321ST
Precio (MXN)
-
Cantidad:
1 Kit
The Invitrogen E-Gel Power Snap Electrophoresis System Starter Kit, SYBR Safe 1.2%, provides everything you need to quickly start using precast 1.2% E-Gel agarose gels with SYBR Safe on the system for a streamlined gel electrophoresis and gel imaging workflow.
The kit includes:
  • 10 E-Gel agarose gels with SYBR Safe, 1% (Cat. No. A42100)
  • E-Gel Power Snap Electrophoresis System (Cat. No. G8300)
  • E-Gel 1 Kb Plus DNA Ladder with E-Gel Sample Loading Buffer (1X) (Cat. No. 10488090)
Para uso exclusivo en investigación. No apto para uso en procedimientos diagnósticos.
Especificaciones
Cantidad1 Kit
Condiciones de envíoTemperatura ambiente
Tensión240 VAC
Tensión máx.240 V CA (fuente de alimentación), 48 V CC (dispositivo de electroforesis)
TipoKit de inicio de sistema de electroforesis
Unit SizeEach
Contenido y almacenamiento
• 1 electrophoresis device
• 1 camera device
• 10 E-Gel agarose gels with SYBR Safe, 1%
• 1 vial E-Gel 1 kb Plus DNA Ladder
• 1 power adapter
• 1 pair of viewing glasses
Store at room temperature.